Search Thermo Fisher Scientific
The Ion AmpliSeq Transcriptome Mouse Gene Expression Kit used with the Ion GeneStudio S5 series systems help to enable fast and affordable gene expression analysis from samples limited in quantity. With simultaneous amplification of more than 20,000 RefSeq genes in a single tube, starting from as little as 10 ng of total RNA, this kit can be an important tool in oncology and translational research.
Figure 1. Correlation of DEGs between Ion AmpliSeq Mouse Gene Expression Kit data and whole transcriptome RNA-Seq data equals 0.943.
The Ion AmpliSeq Transcriptome Mouse Gene Expression Kit offers:
Flyer: Ion AmpliSeq Transcriptome Mouse Gene Expression Kit flyer
Application note: Simple, scalable transcriptome-level gene expression analysis with next-generation sequencing
Learn more about Targeted RNA Sequencing with Ion AmpliSeq RNA panels ›
Transform your research by targeting >20,000 RefSeq transcripts, starting with as little as 10 ng of RNA input and a fast, FFPE-compatible workflow using the Ion AmpliSeq Transcriptome Human Gene Expression Kit and the Ion GeneStudio S5 series sequencing systems.
Flyer: Ion AmpliSeq Transcriptome Human Gene Expression Kit
Application Note: Ion AmpliSeq Transcriptome (a streamlined workflow for gene expression analysis)
Application Note: Ion S5 System (how the system can be used for cancer genomics and transcriptomics research)
Webinar: Application of targeted NGS gene expression-based approach to biomarker discovery
Looking for a gene expression signature in triple-negative breast cancer research samples using the Ion AmpliSeq Transcriptome Human Gene Expression kit
Is there only one way to do RNA-Seq library prep? Hear Milan Radovich, PhD, of Indiana University discuss his research on triple-negative breast cancer expression-based gene signatures.
Quantitative and differential gene expression analysis is fundamental to many biological research applications including gene regulation analysis, correlating gene expression with phenotypic information, and cellular pathway analysis. Conventional gene expression profiling methods include hybridization-based (microarray) and amplification-based (qRT-PCR) technologies. However, methods present certain limitations:
As such, both technologies present workflow challenges when analyzing samples that are limited in quantity or quality, such as degraded RNA from FFPE tissues. Targeted sequencing using NGS and the Ion AmpliSeq Transcriptome Human Gene Expression Kit not only provides a representation of absolute expression, but also can identify and quantify low-abundance transcripts quickly and easily for both intact and degraded samples.
Figure 2. Improved gene-level transcript detection using the Ion AmpliSeq Transcriptome Human Gene Expression Kit and the Ion Proton System. A Venn diagram is shown demonstrating the concordance of genes identified by MAQC microarray data and the Ion AmpliSeq Transcriptome Human Gene Expression Kit.
Amplify >20,000 targets in a single reaction
Products:
Isothermal amplification with Ion Chef System
Products:
Sequencing using Ion S5 System or Ion Proton System
Products:
Amplify >20,000 targets in a single reaction
Products:
Isothermal amplification with Ion Chef System
Products:
Sequencing using Ion S5 System or Ion Proton System
Products:
Adopting NGS in your lab is now simpler than ever before. The Ion GeneStudio S5 series next-generation sequencing systems enable the simplest targeted sequencing workflow for your lab with superior speed and affordability. Focus your research efforts on applications that have the potential to be impactful. These NGS systems have a modular build, so your lab has the flexibility to do multiple targeted sequencing applications on a single system.
Blog Post - Understanding adverse immune response from viral infections
Transcript profiling using the Ion AmpliSeq Transcriptome Human Gene Expression Kit application note
Poster comparing Ion AmpliSeq Transcriptome, RNA-Seq, and real-time PCR data
Application note: Simple, scalable transcriptome-level gene expression analysis with next-generation sequencing