Search Thermo Fisher Scientific
Thermo Scientific single-use cell culture Erlenmeyer flasks are excellent for suspension culture and media preparation. Ready for mixing and storage and featuring crystal-clear polyethylene terephthalate glycol (PETG), these single-use flasks are robust and break resistant.
Standard non-vented closures help prevent spilling and contamination, and optional vented closures (0.22 µm) improves gas exchange, especially beneficial for suspension cell culture.
See how the 5-Liter flask can help save you time, money & space while helping reduce the media and plastics used in a lab all while helping increase efficiency for suspension culturing.
Our angled flask (Cat. No. 4115-5000) has also shown to be the only flask tested capable generating larger protein titers in CHO cells due to its proprietary design (see Figure 1).
Capacity | Outer diameter | Case size |
---|---|---|
125 mL | 66.3 mm | 24 |
250 mL | 82.6 mm | 12 |
500 mL | 101.6 mm | 12 |
1,000 mL | 129.8 mm | 6 |
2,000 mL | 161.5 mm | 4 |
2,800 mL | 203.2 mm | 4 |
5,000 mL | 214 mm | 4 |
Separated below by cap type, sterile single-use shake flasks are available with plain and baffled bottoms. These sterile single-use Erlenmeyer flasks for cell culture are: