Product References
Dlk1 maintains adult mice long-term HSCs by activating Notch signaling to restrict mitochondrial metabolism.
Experimental hematology & oncology
Huang D,Han Y,Tang T,Yang L,Jiang P,Qian W,Zhang Z,Qian X,Zeng X,Qian P
15-5790-82 was used in Flow cytometry/Cell sorting to demonstrate that Dlk1 is essential for maintaining HSC homeostasis, which is realized by governing Notch signalling and restricting mitochondrial metabolic activity.
Wed Jan 18 00:00:00 EST 2023
Nanobodies from camelid mice and llamas neutralize SARS-CoV-2 variants.
Nature
Xu J,Xu K,Jung S,Conte A,Lieberman J,Muecksch F,Lorenzi JCC,Park S,Schmidt F,Wang Z,Huang Y,Luo Y,Nair MS,Wang P,Schulz JE,Tessarollo L,Bylund T,Chuang GY,Olia AS,Stephens T,Teng IT,Tsybovsky Y,Zhou T,Munster V,Ho DD,Hatziioannou T,Bieniasz PD,Nussenzweig MC,Kwong PD,Casellas R
Published figure using IgM monoclonal antibody (Product # 15-5790-82) in Flow Cytometry
Thu Jul 01 00:00:00 EDT 2021
Smc3 dosage regulates B cell transit through germinal centers and restricts their malignant transformation.
Nature immunology
Rivas MA,Meydan C,Chin CR,Challman MF,Kim D,Bhinder B,Kloetgen A,Viny AD,Teater MR,McNally DR,Doane AS,Béguelin W,Fernández MTC,Shen H,Wang X,Levine RL,Chen Z,Tsirigos A,Elemento O,Mason CE,Melnick AM
Published figure using IgM monoclonal antibody (Product # 15-5790-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2021
A platform-agnostic, function first-based antibody discovery strategy using plasmid-free mammalian expression of antibodies.
mAbs
Zhang R,Prabakaran P,Yu X,Mackness BC,Boudanova E,Hopke J,Sancho J,Saleh J,Cho H,Zhang N,Simonds-Mannes H,Stimple SD,Hoffmann D,Park A,Chowdhury PS,Rao SP
Published figure using IgM monoclonal antibody (Product # 15-5790-82) in Flow Cytometry
Wed Jan 19 00:00:00 EST 2022
Construction, characterization, and immunization of nanoparticles that display a diverse array of influenza HA trimers.
PloS one
Cohen AA,Yang Z,Gnanapragasam PNP,Ou S,Dam KA,Wang H,Bjorkman PJ
Published figure using IgM monoclonal antibody (Product # 15-5790-82) in Flow Cytometry
Fri Oct 15 00:00:00 EDT 2021
B Cell Diversification Is Uncoupled from SAP-Mediated Selection Forces in Chronic Germinal Centers within Peyer's Patches.
Cell reports
Biram A,Winter E,Denton AE,Zaretsky I,Dassa B,Bemark M,Linterman MA,Yaari G,Shulman Z
Published figure using IgM monoclonal antibody (Product # 15-5790-82) in Flow Cytometry
Tue Feb 11 00:00:00 EST 2020
Chronic Viral Infection Promotes Efficient Germinal Center B Cell Responses.
Cell reports
Fallet B,Hao Y,Florova M,Cornille K,de Los Aires AV,Girelli Zubani G,Ertuna YI,Greiff V,Menzel U,Hammad K,Merkler D,Reddy ST,Weill JC,Reynaud CA,Pinschewer DD
Published figure using IgM monoclonal antibody (Product # 15-5790-82) in Flow Cytometry
Tue Jan 28 00:00:00 EST 2020
TET enzymes control antibody production and shape the mutational landscape in germinal centre B cells.
The FEBS journal
Schoeler K,Aufschnaiter A,Messner S,Derudder E,Herzog S,Villunger A,Rajewsky K,Labi V
Published figure using IgM monoclonal antibody (Product # 15-5790-82) in Flow Cytometry
Sun Sep 01 00:00:00 EDT 2019
N-Cadherin-Expressing Bone and Marrow Stromal Progenitor Cells Maintain Reserve Hematopoietic Stem Cells.
Cell reports
Zhao M,Tao F,Venkatraman A,Li Z,Smith SE,Unruh J,Chen S,Ward C,Qian P,Perry JM,Marshall H,Wang J,He XC,Li L
15-5790 was used in Flow cytometry/Cell sorting to report that N-cad+ cells were functional bone and marrow stromal progenitor cells, giving rise to osteoblasts, adipocytes, and chondrocytes both in vitro and in vivo.
Tue Jan 15 00:00:00 EST 2019
A protocol for generating induced T cells by reprogramming B cells in vivo.
Cell regeneration (London, England)
Weng Q,Hu F,Zhang M,Dong Y,Lv C,Wang Y,Liu X,Wang J
Published figure using IgM monoclonal antibody (Product # 15-5790-82) in Flow Cytometry
Sat Sep 01 00:00:00 EDT 2018
Retinoid-Sensitive Epigenetic Regulation of the Hoxb Cluster Maintains Normal Hematopoiesis and Inhibits Leukemogenesis.
Cell stem cell
Qian P,De Kumar B,He XC,Nolte C,Gogol M,Ahn Y,Chen S,Li Z,Xu H,Perry JM,Hu D,Tao F,Zhao M,Han Y,Hall K,Peak A,Paulson A,Zhao C,Venkatraman A,Box A,Perera A,Haug JS,Parmely T,Li H,Krumlauf R,Li L
15-5790 was used in Flow cytometry/Cell sorting to demonstrate roles for retinoid signaling and the DERARE in maintaining HSCs and preventing leukemogenesis by coordinate regulation of Hoxb genes.
Thu May 03 00:00:00 EDT 2018
Conversion of T cells to B cells by inactivation of polycomb-mediated epigenetic suppression of the B-lineage program.
Genes & development
Ikawa T,Masuda K,Endo TA,Endo M,Isono K,Koseki Y,Nakagawa R,Kometani K,Takano J,Agata Y,Katsura Y,Kurosaki T,Vidal M,Koseki H,Kawamoto H
Published figure using IgM monoclonal antibody (Product # 15-5790-82) in Flow Cytometry
Tue Nov 15 00:00:00 EST 2016
Ligase I and ligase III mediate the DNA double-strand break ligation in alternative end-joining.
Proceedings of the National Academy of Sciences of the United States of America
Lu G,Duan J,Shu S,Wang X,Gao L,Guo J,Zhang Y
Published figure using IgM monoclonal antibody (Product # 15-5790-82) in Flow Cytometry
Tue Feb 02 00:00:00 EST 2016
Editing of mouse and human immunoglobulin genes by CRISPR-Cas9 system.
Nature communications
Cheong TC,Compagno M,Chiarle R
Published figure using IgM monoclonal antibody (Product # 15-5790-82) in Flow Cytometry
Wed Mar 09 00:00:00 EST 2016
Distinctive Mesenchymal-Parenchymal Cell Pairings Govern B Cell Differentiation in the Bone Marrow.
Stem cell reports
Yu VW,Lymperi S,Oki T,Jones A,Swiatek P,Vasic R,Ferraro F,Scadden DT
15-5790 was used in Flow cytometry/Cell sorting to study the notion that bone marrow is a composite of specialised niches, providing distinctive functional units to regulate hematopoiesis.
Tue Aug 09 00:00:00 EDT 2016
B cell autophagy mediates TLR7-dependent autoimmunity and inflammation.
Autophagy
Weindel CG,Richey LJ,Bolland S,Mehta AJ,Kearney JF,Huber BT
Published figure using IgM monoclonal antibody (Product # 15-5790-82) in Flow Cytometry
Mon Apr 18 00:00:00 EDT 2016
Mitochondrial ATP transporter Ant2 depletion impairs erythropoiesis and B lymphopoiesis.
Cell death and differentiation
Cho J,Seo J,Lim CH,Yang L,Shiratsuchi T,Lee MH,Chowdhury RR,Kasahara H,Kim JS,Oh SP,Lee YJ,Terada N
Published figure using IgM monoclonal antibody (Product # 15-5790-82) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2015
Meningeal Infiltration of the Spinal Cord by Non-Classically Activated B Cells is Associated with Chronic Disease Course in a Spontaneous B Cell-Dependent Model of CNS Autoimmune Disease.
Frontiers in immunology
Dang AK,Tesfagiorgis Y,Jain RW,Craig HC,Kerfoot SM
15-5790 was used in Flow cytometry/Cell sorting to characterise B cell infiltration of the spinal cord in a B cell-dependent spontaneous model of CNS autoimmunity, showing meningeal infiltration of the spinal cord by non-classically activated B cells.
Wed Oct 07 00:00:00 EDT 2015
VprBP Is Required for Efficient Editing and Selection of Igκ+ B Cells, but Is Dispensable for Igλ+ and Marginal Zone B Cell Maturation and Selection.
Journal of immunology (Baltimore, Md. : 1950)
Palmer VL,Aziz-Seible R,Kassmeier MD,Rothermund M,Perry GA,Swanson PC
15-5790 was used in Flow cytometry/Cell sorting to determine the contribution of the protein, VprBP, to B cell development and selection.
Sat Aug 15 00:00:00 EDT 2015
The proximal J kappa germline-transcript promoter facilitates receptor editing through control of ordered recombination.
PloS one
Vettermann C,Timblin GA,Lim V,Lai EC,Schlissel MS
15-5790 was used in Flow cytometry/Cell sorting to identify the proximal GT promoter as the first cis-element known to regulate the internal order of Jκ rearrangements.
Tue Jan 26 00:00:00 EST 2016
Elucidation of the enigmatic IgD class-switch recombination via germline deletion of the IgH 3' regulatory region.
The Journal of experimental medicine
Rouaud P,Saintamand A,Saad F,Carrion C,Lecardeur S,Cogné M,Denizot Y
Published figure using IgM monoclonal antibody (Product # 15-5790-82) in Flow Cytometry
Mon May 05 00:00:00 EDT 2014
A component of the mir-17-92 polycistronic oncomir promotes oncogene-dependent apoptosis.
eLife
Olive V,Sabio E,Bennett MJ,De Jong CS,Biton A,McGann JC,Greaney SK,Sodir NM,Zhou AY,Balakrishnan A,Foth M,Luftig MA,Goga A,Speed TP,Xuan Z,Evan GI,Wan Y,Minella AC,He L
Published figure using IgM monoclonal antibody (Product # 15-5790-82) in Flow Cytometry
Tue Oct 15 00:00:00 EDT 2013
A mouse model for inducible overexpression of Prdm14 results in rapid-onset and highly penetrant T-cell acute lymphoblastic leukemia (T-ALL).
Disease models & mechanisms
Carofino BL,Ayanga B,Justice MJ
Published figure using IgM monoclonal antibody (Product # 15-5790-82) in Flow Cytometry
Fri Nov 01 00:00:00 EDT 2013
Galectin 1 modulates plasma cell homeostasis and regulates the humoral immune response.
Journal of immunology (Baltimore, Md. : 1950)
Anginot A,Espeli M,Chasson L,Mancini SJ,Schiff C
15-5790 was used in Flow cytometry/Cell sorting to elucidate the pattern of GAL1 expression and the function of GAL1 in B cells.
Sat Jun 01 00:00:00 EDT 2013
Critical role of B cell lymphoma 10 in BAFF-regulated NF-κB activation and survival of anergic B cells.
Journal of immunology (Baltimore, Md. : 1950)
Yu M,Chen Y,He Y,Podd A,Fu G,Wright JA,Kleiman E,Khan WN,Wen R,Wang D
Published figure using IgM monoclonal antibody (Product # 15-5790-82) in Flow Cytometry
Sat Dec 01 00:00:00 EST 2012
Nuclear export of the NF-κB inhibitor IκBα is required for proper B cell and secondary lymphoid tissue formation.
Immunity
Wuerzberger-Davis SM,Chen Y,Yang DT,Kearns JD,Bates PW,Lynch C,Ladell NC,Yu M,Podd A,Zeng H,Huang TT,Wen R,Hoffmann A,Wang D,Miyamoto S
Published figure using IgM monoclonal antibody (Product # 15-5790-82) in Flow Cytometry
Fri Feb 25 00:00:00 EST 2011
Deletion of genes encoding PU.1 and Spi-B in B cells impairs differentiation and induces pre-B cell acute lymphoblastic leukemia.
Blood
Sokalski KM,Li SK,Welch I,Cadieux-Pitre HA,Gruca MR,DeKoter RP
15-5790 was used in Flow cytometry/Cell sorting to investigate the impairment of differentiation of B cells and induction of pre-B cell acute lymphoblastic leukemia through the deletion of genes encoding PU.1 and Spi-1 in B cells.
Thu Sep 08 00:00:00 EDT 2011
Regulation of follicular B cell differentiation by the related E26 transformation-specific transcription factors PU.1, Spi-B, and Spi-C.
Journal of immunology (Baltimore, Md. : 1950)
DeKoter RP,Geadah M,Khoosal S,Xu LS,Thillainadesan G,Torchia J,Chin SS,Garrett-Sinha LA
15-5790 was used in Flow cytometry/Cell sorting to demonstrate the role of the E26 transformation-specific transcription factors PU.1, Spi-B, and Spi-C in follicular B cell development.
Wed Dec 15 00:00:00 EST 2010
A critical role of TAK1 in B-cell receptor-mediated nuclear factor kappaB activation.
Blood
Schuman J,Chen Y,Podd A,Yu M,Liu HH,Wen R,Chen ZJ,Wang D
Published figure using IgM monoclonal antibody (Product # 15-5790-82) in Flow Cytometry
Thu May 07 00:00:00 EDT 2009
miR-19 is a key oncogenic component of mir-17-92.
Genes & development
Olive V,Bennett MJ,Walker JC,Ma C,Jiang I,Cordon-Cardo C,Li QJ,Lowe SW,Hannon GJ,He L
Published figure using IgM monoclonal antibody (Product # 15-5790-82) in Flow Cytometry
Tue Dec 15 00:00:00 EST 2009
Phospholipase Cgamma2 contributes to light-chain gene activation and receptor editing.
Molecular and cellular biology
Bai L,Chen Y,He Y,Dai X,Lin X,Wen R,Wang D
Published figure using IgM monoclonal antibody (Product # 15-5790-82) in Flow Cytometry
Sat Sep 01 00:00:00 EDT 2007