Product References
TGFβ1+CCR5+ neutrophil subset increases in bone marrow and causes age-related osteoporosis in male mice.
Nature communications
Li J,Yao Z,Liu X,Duan R,Yi X,Ayoub A,Sanders JO,Mesfin A,Xing L,Boyce BF
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Wed Jan 11 00:00:00 EST 2023
Sympathetic Nerves Coordinate Corneal Epithelial Wound Healing by Controlling the Mobilization of Ly6Chi Monocytes From the Spleen to the Injured Cornea.
Investigative ophthalmology & visual science
He S,Liu J,Xue Y,Fu T,Li Z
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Fri Sep 01 00:00:00 EDT 2023
Macrophagic AMPKα1 orchestrates regenerative inflammation induced by glucocorticoids.
EMBO reports
Caratti G,Desgeorges T,Juban G,Stifel U,Fessard A,Koenen M,Caratti B,Théret M,Skurk C,Chazaud B,Tuckermann JP,Mounier R
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Mon Feb 06 00:00:00 EST 2023
Group 2 Innate Lymphoid Cells Protect Mice from Abdominal Aortic Aneurysm Formation via IL5 and Eosinophils.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zhang Y,Liu T,Deng Z,Fang W,Zhang X,Zhang S,Wang M,Luo S,Meng Z,Liu J,Sukhova GK,Li D,McKenzie ANJ,Libby P,Shi GP,Guo J
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2023
The NKCC1 ion transporter modulates microglial phenotype and inflammatory response to brain injury in a cell-autonomous manner.
PLoS biology
Tóth K,Lénárt N,Berki P,Fekete R,Szabadits E,Pósfai B,Cserép C,Alatshan A,Benkő S,Kiss D,Hübner CA,Gulyás A,Kaila K,Környei Z,Dénes Á
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Sat Jan 01 00:00:00 EST 2022
Gene-dosage effect of Pfkfb3 on monocyte/macrophage biology in atherosclerosis.
British journal of pharmacology
Guo S,Li A,Fu X,Li Z,Cao K,Song M,Huang S,Li Z,Yan J,Wang L,Dai X,Feng D,Wang Y,He J,Huo Y,Xu Y
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Tue Nov 01 00:00:00 EDT 2022
Mineralocorticoid receptor antagonists and glucocorticoids differentially affect skeletal muscle inflammation and pathology in muscular dystrophy.
JCI insight
Howard ZM,Gomatam CK,Rabolli CP,Lowe J,Piepho AB,Bansal SS,Accornero F,Rafael-Fortney JA
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Mon Oct 10 00:00:00 EDT 2022
Therapeutic CFTR Correction Normalizes Systemic and Lung-Specific S1P Level Alterations Associated with Heart Failure.
International journal of molecular sciences
Uhl FE,Vanherle L,Matthes F,Meissner A
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Fri Jan 14 00:00:00 EST 2022
Chronic T cell proliferation in brains after stroke could interfere with the efficacy of immunotherapies.
The Journal of experimental medicine
Heindl S,Ricci A,Carofiglio O,Zhou Q,Arzberger T,Lenart N,Franzmeier N,Hortobagyi T,Nelson PT,Stowe AM,Denes A,Edbauer D,Liesz A
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Mon Aug 02 00:00:00 EDT 2021
Pancreatic Cancer Chemotherapy Is Potentiated by Induction of Tertiary Lymphoid Structures in Mice.
Cellular and molecular gastroenterology and hepatology
Delvecchio FR,Fincham REA,Spear S,Clear A,Roy-Luzarraga M,Balkwill FR,Gribben JG,Bombardieri M,Hodivala-Dilke K,Capasso M,Kocher HM
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Mon Mar 28 00:00:00 EDT 2022
Disrupted Iron Metabolism and Mortality during Co-infection with Malaria and an Intestinal Gram-Negative Extracellular Pathogen.
Cell reports
Dos Santos LI,Torres TA,Diniz SQ,Gonçalves R,Caballero-Flores G,Núñez G,Gazzinelli RT,Maloy KJ,Ribeiro do V Antonelli L
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Tue Jan 12 00:00:00 EST 2021
Myeloid DJ-1 deficiency protects acetaminophen-induced acute liver injury through decreasing inflammatory response.
Aging
Wang B,Li J,Jiao J,Xu M,Luo Y,Wang F,Xia Q,Gao Y,Feng Y,Kong X,Sun X
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Wed Jul 21 00:00:00 EDT 2021
NOX4 regulates macrophage apoptosis resistance to induce fibrotic progression.
The Journal of biological chemistry
Larson-Casey JL,Gu L,Kang J,Dhyani A,Carter AB
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Thu Jul 01 00:00:00 EDT 2021
STING Activated Tumor-Intrinsic Type I Interferon Signaling Promotes CXCR3 Dependent Antitumor Immunity in Pancreatic Cancer.
Cellular and molecular gastroenterology and hepatology
Vonderhaar EP,Barnekow NS,McAllister D,McOlash L,Eid MA,Riese MJ,Tarakanova VL,Johnson BD,Dwinell MB
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Mon Jan 24 00:00:00 EST 2022
Abraxane-induced bone marrow CD11b+ myeloid cell depletion in tumor-bearing mice is visualized by μPET-CT with 64Cu-labeled anti-CD11b and prevented by anti-CSF-1.
Theranostics
Cao Q,Huang Q,Wang YA,Li C
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Fri Jul 23 00:00:00 EDT 2021
Inflammatory resolution and vascular barrier restoration after retinal ischemia reperfusion injury.
Journal of neuroinflammation
Abcouwer SF,Shanmugam S,Muthusamy A,Lin CM,Kong D,Hager H,Liu X,Antonetti DA
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Thu Aug 26 00:00:00 EDT 2021
Roles of lung-recruited monocytes and pulmonary Vascular Endothelial Growth Factor (VEGF) in resolving Ventilator-Induced Lung Injury (VILI).
PloS one
Lin CK,Huang TH,Yang CT,Shi CS
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Tue Oct 12 00:00:00 EDT 2021
Plaque-associated myeloid cells derive from resident microglia in an Alzheimer's disease model.
The Journal of experimental medicine
Reed-Geaghan EG,Croxford AL,Becher B,Landreth GE
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Mon Apr 06 00:00:00 EDT 2020
Proenkephalin+ regulatory T cells expanded by ultraviolet B exposure maintain skin homeostasis with a healing function.
Proceedings of the National Academy of Sciences of the United States of America
Shime H,Odanaka M,Tsuiji M,Matoba T,Imai M,Yasumizu Y,Uraki R,Minohara K,Watanabe M,Bonito AJ,Fukuyama H,Ohkura N,Sakaguchi S,Morita A,Yamazaki S
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Tue Aug 25 00:00:00 EDT 2020
IgE Contributes to Atherosclerosis and Obesity by Affecting Macrophage Polarization, Macrophage Protein Network, and Foam Cell Formation.
Arteriosclerosis, thrombosis, and vascular biology
Zhang X,Li J,Luo S,Wang M,Huang Q,Deng Z,de Febbo C,Daoui A,Liew PX,Sukhova GK,Metso J,Jauhiainen M,Shi GP,Guo J
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Sun Mar 01 00:00:00 EST 2020
Modulation of the functions of myeloid-derived suppressor cells : a new strategy of hydrogen sulfide anti-cancer effects.
British journal of pharmacology
De Cicco P,Ercolano G,Rubino V,Terrazzano G,Ruggiero G,Cirino G,Ianaro A
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Sat Feb 01 00:00:00 EST 2020
Temporospatial distribution and transcriptional profile of retinal microglia in the oxygen-induced retinopathy mouse model.
Glia
Boeck M,Thien A,Wolf J,Hagemeyer N,Laich Y,Yusuf D,Backofen R,Zhang P,Boneva S,Stahl A,Hilgendorf I,Agostini H,Prinz M,Wieghofer P,Schlunck G,Schlecht A,Lange C
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2020
Characterization and regulation of osteoclast precursors following chronic Porphyromonas gingivalis infection.
Journal of leukocyte biology
Zhao Y,Li Z,Su L,Ballesteros-Tato A,Katz J,Michalek SM,Feng X,Zhang P
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Thu Oct 01 00:00:00 EDT 2020
Antitumor Effect and Immune Response of Nanosecond Pulsed Electric Fields in Pancreatic Cancer.
Frontiers in oncology
Zhao J,Chen S,Zhu L,Zhang L,Liu J,Xu D,Tian G,Jiang T
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Fri Oct 28 00:00:00 EDT 2022
Effects of Hypertonic Saline and Hydroxyethyl Starch on Myeloid-Derived Suppressor Cells in Hemorrhagic Shock Mice under Secondary Bacterial Attack.
BioMed research international
Jiang JK,Hong LJ,Lu YQ
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Wed Dec 16 00:00:00 EST 2020
Site-1 protease ablation in the osterix-lineage in mice results in bone marrow neutrophilia and hematopoietic stem cell alterations.
Biology open
Patra D,Kim J,Zhang Q,Tycksen E,Sandell LJ
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Tue Jun 23 00:00:00 EDT 2020
Site-1 protease ablation in the osterix-lineage in mice results in bone marrow neutrophilia and hematopoietic stem cell alterations.
Biology open
Patra D,Kim J,Zhang Q,Tycksen E,Sandell LJ
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Tue Jun 23 00:00:00 EDT 2020
The impact of Clonorchis sinensis infection on immune response in mice with type II collagen-induced arthritis.
BMC immunology
Li X,Yang Y,Qin S,Kong F,Yan C,Cheng W,Pan W,Yu Q,Hua H,Zheng K,Tang R
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Mon Feb 17 00:00:00 EST 2020
GDF3 Protects Mice against Sepsis-Induced Cardiac Dysfunction and Mortality by Suppression of Macrophage Pro-Inflammatory Phenotype.
Cells
Wang L,Li Y,Wang X,Wang P,Essandoh K,Cui S,Huang W,Mu X,Liu Z,Wang Y,Peng T,Fan GC
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Fri Jan 03 00:00:00 EST 2020
The Transcription Factor Nfix Requires RhoA-ROCK1 Dependent Phagocytosis to Mediate Macrophage Skewing during Skeletal Muscle Regeneration.
Cells
Saclier M,Lapi M,Bonfanti C,Rossi G,Antonini S,Messina G
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Fri Mar 13 00:00:00 EDT 2020
Extracellular miR-146a-5p Induces Cardiac Innate Immune Response and Cardiomyocyte Dysfunction.
ImmunoHorizons
Shimada BK,Yang Y,Zhu J,Wang S,Suen A,Kronstadt SM,Jeyaram A,Jay SM,Zou L,Chao W
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Mon Sep 21 00:00:00 EDT 2020
Resident macrophages acquire innate immune memory in staphylococcal skin infection.
eLife
Feuerstein R,Forde AJ,Lohrmann F,Kolter J,Ramirez NJ,Zimmermann J,Gomez de Agüero M,Henneke P
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Wed Jul 08 00:00:00 EDT 2020
Squalene emulsion-based vaccine adjuvants stimulate CD8 T cell, but not antibody responses, through a RIPK3-dependent pathway.
eLife
Kim EH,Woodruff MC,Grigoryan L,Maier B,Lee SH,Mandal P,Cortese M,Natrajan MS,Ravindran R,Ma H,Merad M,Gitlin AD,Mocarski ES,Jacob J,Pulendran B
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Tue Jun 09 00:00:00 EDT 2020
Gasdermin D in macrophages restrains colitis by controlling cGAS-mediated inflammation.
Science advances
Ma C,Yang D,Wang B,Wu C,Wu Y,Li S,Liu X,Lassen K,Dai L,Yang S
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Fri May 01 00:00:00 EDT 2020
Host Cathelicidin Exacerbates Group B Streptococcus Urinary Tract Infection.
mSphere
Patras KA,Coady A,Babu P,Shing SR,Ha AD,Rooholfada E,Brandt SL,Geriak M,Gallo RL,Nizet V
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Wed Apr 22 00:00:00 EDT 2020
Increased flux through the mevalonate pathway mediates fibrotic repair without injury.
The Journal of clinical investigation
Larson-Casey JL,Vaid M,Gu L,He C,Cai GQ,Ding Q,Davis D,Berryhill TF,Wilson LS,Barnes S,Neighbors JD,Hohl RJ,Zimmerman KA,Yoder BK,Longhini ALF,Hanumanthu VS,Surolia R,Antony VB,Carter AB
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Fri Nov 01 00:00:00 EDT 2019
Regulatory T Cells Control the Switch From in situ to Invasive Breast Cancer.
Frontiers in immunology
Martinez LM,Robila V,Clark NM,Du W,Idowu MO,Rutkowski MR,Bos PD
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Tue Sep 29 00:00:00 EDT 2020
Differentiated macrophages acquire a pro-inflammatory and cell death-resistant phenotype due to increasing XIAP and p38-mediated inhibition of RipK1.
The Journal of biological chemistry
Rijal D,Ariana A,Wight A,Kim K,Alturki NA,Aamir Z,Ametepe ES,Korneluk RG,Tiedje C,Menon MB,Gaestel M,McComb S,Sad S
Published figure using Ly-6C monoclonal antibody (Product # 62-5932-82) in Flow Cytometry
Fri Jul 27 00:00:00 EDT 2018