Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Thermo Fisher Scientific provides engineers and scientists a wide range of sensors for the measurement of strain, acoustics, liquid level, and position from leading global manufacturers such as Micro-Measurements and MTS Sensors.
We have a wide range of non-contact linear position sensors from MTS Sensors for industrial machinery and light industrial applications, along with position sensors for mobile machinery which are specifically designed for direct stroke measurement in hydraulic cylinders.
Our selection of precision liquid level sensors and transmitters measure multiple process variables from one sensor for lower installed costs compared to alternative level technologies. Level Plus liquid level transmitters are designed for automatic tank gauging of aboveground and underground storage tanks. Level Plus transmitters feature 3-in-1 measurement of the product level, interface, and temperature through a single tank opening.