Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Thermo Fisher Scientific offers a wide range of non-contact linear position sensors from MTS Sensors. The Temposonics line of position sensors are designed for use in industrial applications and in mobile machinery. These devices are able to adapt from simple, standard measurements to tasks that require highly precise information.
Temposonics position sensors for industrial applications are developed for external mounting or in-cylinder applications where the electronics can be external. Stroke lengths are available from 25 mm to 20 m. The sensors offer analogue output or dual output (position and velocity) with SSI, DeviceNet, CANbus, Profibus, Ethernet IP, and Ethercat interface. High quality displacement and velocity signals enable superior control. Different sensor models and options (e.g. multi-position measurement, redundant sensors, ATEX-approval) meet every need, from high precision measurement tasks to simple standard applications.
Temposonics® R-Series V Position Sensors | Temposonics® R-Series Position Sensors | Temposonics® G-Series Position Sensors |
---|---|---|
The MTS Sensors R-Series V position sensors are the successors to the current fourth generation. The new sensors have higher resistance to vibration and high temperatures and are ready for Industry 4.0, fitting perfectly into existing applications. | The MTS Sensors Temposonics R-Series position sensors offer optimum performance for industrial applications. | G-Series sensors feature a microprocessor-based design with enhanced diagnostics and programmability, offering the flexibility to fit a wide range of applications. |
Temposonics position sensors for mobile machinery are specifically designed for direct stroke measurement in hydraulic cylinders. They increase the machine’s productivity, automate recurring operation sequences, and reduce maintenance and downtime. Even in harsh environments, at high temperature or high pressure, these sensors work reliably and are maintenance-free. Stroke lengths range from 50 mm to 5000 mm, and outputs include CANbus, J1939, scaled PWM, and analogue interfaces. Redundant models or SIL2 sensors grant maximum safety for critical applications.
MH-Series In-Cylinder Linear Position Sensors | FMH In-Cylinder Linear Position Sensor | MXR Analogue Output Position Sensor |
---|---|---|
MTS Sensors Temposonics MH Series model MH magnetostrictive position sensors measure the absolute position of hydraulic cylinders. Model MH mobile hydraulic sensors can be fully sealed and embedded in a cylinder, providing excellent protection against the environment and EMI and ensures a long operating life. | MTS Sensors Temposonics MH Series model FMH magnetostrictive position sensors are designed for use with hydraulic cylinders in mobile applications. The externally threaded Flexible MH sensor features an innovative two-part design. | The MTS Sensors Temposonics MH Series Model MXR magnetostrictive position sensor measures the absolute position of linear displacements. The Model MXR can be mounted externally to a hydraulic cylinder or between two linkage points. |
Temposonics position sensors for industrial applications are developed for external mounting or in-cylinder applications where the electronics can be external. Stroke lengths are available from 25 mm to 20 m. The sensors offer analogue output or dual output (position and velocity) with SSI, DeviceNet, CANbus, Profibus, Ethernet IP, and Ethercat interface. High quality displacement and velocity signals enable superior control. Different sensor models and options (e.g. multi-position measurement, redundant sensors, ATEX-approval) meet every need, from high precision measurement tasks to simple standard applications.
Temposonics® R-Series V Position Sensors | Temposonics® R-Series Position Sensors | Temposonics® G-Series Position Sensors |
---|---|---|
The MTS Sensors R-Series V position sensors are the successors to the current fourth generation. The new sensors have higher resistance to vibration and high temperatures and are ready for Industry 4.0, fitting perfectly into existing applications. | The MTS Sensors Temposonics R-Series position sensors offer optimum performance for industrial applications. | G-Series sensors feature a microprocessor-based design with enhanced diagnostics and programmability, offering the flexibility to fit a wide range of applications. |
Temposonics position sensors for mobile machinery are specifically designed for direct stroke measurement in hydraulic cylinders. They increase the machine’s productivity, automate recurring operation sequences, and reduce maintenance and downtime. Even in harsh environments, at high temperature or high pressure, these sensors work reliably and are maintenance-free. Stroke lengths range from 50 mm to 5000 mm, and outputs include CANbus, J1939, scaled PWM, and analogue interfaces. Redundant models or SIL2 sensors grant maximum safety for critical applications.
MH-Series In-Cylinder Linear Position Sensors | FMH In-Cylinder Linear Position Sensor | MXR Analogue Output Position Sensor |
---|---|---|
MTS Sensors Temposonics MH Series model MH magnetostrictive position sensors measure the absolute position of hydraulic cylinders. Model MH mobile hydraulic sensors can be fully sealed and embedded in a cylinder, providing excellent protection against the environment and EMI and ensures a long operating life. | MTS Sensors Temposonics MH Series model FMH magnetostrictive position sensors are designed for use with hydraulic cylinders in mobile applications. The externally threaded Flexible MH sensor features an innovative two-part design. | The MTS Sensors Temposonics MH Series Model MXR magnetostrictive position sensor measures the absolute position of linear displacements. The Model MXR can be mounted externally to a hydraulic cylinder or between two linkage points. |