Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Thermo Fisher Scientific offers a wide range of precision liquid level sensors and transmitters. Level Plus liquid level transmitters are designed for automatic tank gauging of aboveground and underground storage tanks. Level Plus transmitters feature 3-in-1 measurement of the product level, interface, and temperature through a single tank opening. Measuring multiple process variables from one sensor provides for lower installed costs compared to alternative level technologies.
Our selection of Level Plus ® liquid level transmitters address your various application needs. The SoClean®, Tank Slayer®, CHAMBERED, and RefineME® liquid level transmitters all satisfy the demand for an accurate and robust liquid-level sensor with unsurpassed flexibility to meet most process application conditions. Each model comes with unique features suited to the individual challenges presented by various working environments. Once the transmitter is installed and calibrated, there is no requirement for scheduled maintenance or recalibration. Additionally, the Level Plus® CHAMBERED liquid level transmitter (not pictured below) can be paired with a Magnetic Level Gauge to provide external measurement of most Magnetic Level Gauges (MLG) from popular suppliers.
SoClean® Liquid Level Transmitter | Tank Slayer® Liquid Level Transmitter | RefineME® Liquid Level Transmitter |
---|---|---|
SoClean® from MTS Sensors is a sanitary level transmitter for Automatic Tank Gauging for the pharmaceutical and food and beverage industries. It provides 4-in-1 measurement using one process opening for product level, interface level, and temperature measurements. | Tank Slayer® from MTS Sensors is a flexible level transmitter for Automatic Tank Gauging of bulk Storage tanks for inventory monitoring. It provides 4-in-1 measurement using one process opening for product level, interface level, and temperature measurements. | RefineME® from MTS Sensors is an industrial rigid level transmitter for Automatic Tank Gauging of process tanks in harsh environments at refineries, chemical plants, and mines. It provides 4-in-1 measurement using one process opening for product level, interface level, and temperature measurements. |