Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Fluorophore Selection Guide |
Fluorophores enable researchers to expand the limits of their multicolor experiments in high content analysis, microscopy, and flow cytometry platforms. Our fluorescent dyes are offered conjugated to antibodies, streptavidin, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection. They are also available as labeling reagents and kits allowing researchers to create labeled biomolecules for use in various applications as well as for probing biological structures, functions, and interactions. You can find information about each fluorophore by browsing the tables below.
On this page, you can also find fluorophore brands to explore our different dye brands and resources to explore protocols and selection guide posters for flow cytometry and imaging.
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
Brilliant Ultra Violet 395 | 347 | 399 | Polymer dye | flow cytometry |
Alexa Fluor 350 | 343 | 441 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Brilliant Ultra Violet 496 | 348 | 493 | Tandem polymer dye | flow cytometry |
Qdot 525 Probe | 300 | 525 | Quantum dot | microscopy; flow cytometry |
Brilliant Ultra Violet 563 | 349 | 563 | Tandem polymer dye | flow cytometry |
Qdot 565 Probe | 300 | 564 | Quantum dot | microscopy; flow cytometry |
Qdot 605 Probe | 300 | 603 | Quantum dot | microscopy; flow cytometry |
Brilliant Ultra Violet 615 | 350 | 607 | Tandem polymer dye | flow cytometry |
Qdot 655 Probe | 300 | 654 | Quantum dot | microscopy; flow cytometry |
Brilliant Ultra Violet 661 | 349 | 659 | Tandem polymer dye | flow cytometry |
Qdot 705 Probe | 300 | 702 | Quantum dot | microscopy; flow cytometry |
Brilliant Ultra Violet 737 | 350 | 740 | Tandem polymer dye | flow cytometry |
Qdot 800 Probe | 300 | 792 | Quantum dot | microscopy; flow cytometry |
Brilliant Ultra Violet 805 | 349 | 804 | Tandem polymer dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
Alexa Fluor 405 | 401 | 422 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Brilliant Violet 421 | 406 | 423 | Polymer dye | flow cytometry |
Super Bright 436 | 413 | 431 | Polymer dye | flow cytometry |
eFluor 450 | 405 | 445 | Small organic dye | flow cytometry |
Pacific Blue | 404 | 455 | Small organic dye | flow cytometry |
Coumarin and Coumarin Derivatives | 387 | 470 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Brilliant Violet 480 | 437 | 478 | Polymer dye | flow cytometry |
Cyan Fluorescent Protein (CFP) | 435 | 485 | Expressed fluorescent protein | microscopy; flow cytometry |
eFluor 506 | 419 | 508 | Small organic dye | flow cytometry |
Pacific Green | 411 | 510 | Small organic dye | flow cytometry |
Pacific Orange | 400 | 551 | Small organic dye | flow cytometry |
Super Bright 600 | 414 | 601 | Tandem polymer dye | flow cytometry |
Brilliant Violet 605 | 405 | 605 | Tandem polymer dye | flow cytometry |
Super Bright 645 | 414 | 645 | Tandem polymer dye | flow cytometry |
Brilliant Violet 650 | 407 | 646 | Tandem polymer dye | flow cytometry |
Super Bright 702 | 413 | 702 | Tandem polymer dye | flow cytometry |
Brilliant Violet 711 | 406 | 711 | Tandem polymer dye | flow cytometry |
Super Bright 780 | 413 | 780 | Tandem polymer dye | flow cytometry |
Brilliant Violet 786 | 406 | 788 | Tandem polymer dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
Green Fluorescent Protein (GFP) | 488 | 510 | Expressed fluorescent protein | microscopy; flow cytometry |
BODIPY FL | 502 | 511 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Blue 510 | 493 | 513 | Phiton-based DNA scaffold dye | flow cytometry |
Fluorescein (FITC) | 498 | 517 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Alexa Fluor 488 | 499 | 520 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Oregon Green 488 | 498 | 526 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Blue 530 | 508 | 527 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Blue 555 | 493 | 555 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Blue 585 | 493 | 586 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Blue 610-30S | 508 | 615 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Blue 610-70S | 492 | 616 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Blue 660-40S | 508 | 664 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Blue 660-120S | 492 | 665 | Phiton-based DNA scaffold dye | flow cytometry |
PerCP-Cyanine5.5 | 489 | 679 | Protein-based tandem dye | flow cytometry |
PerCP-eFluor 710 | 482 | 708 | Protein-based tandem dye | flow cytometry |
NovaFluor Blue 725 | 492 | 725 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Blue 760 | 490 | 764 | Phiton-based DNA scaffold dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
Alexa Fluor 532 | 534 | 553 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Cy3 | 554 | 566 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Yellow 570 | 551 | 566 | Phiton-based DNA scaffold dye | flow cytometry |
Alexa Fluor 555 | 553 | 568 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Alexa Fluor 561 | 556 | 569 | Small organic dye | flow cytometry |
Alexa Fluor 546 | 561 | 572 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
R-phycoerythrin (R-PE) | 565 | 576 | Protein-based fluorophore | flow cytometry |
Tetramethylrhodamine (TRITC) | 552 | 578 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Red Fluorescent Protein (RFP) | 555 | 584 | Expressed fluorescent protein | microscopy; flow cytometry |
NovaFluor Yellow 590 | 563 | 594 | Phiton-based DNA scaffold dye | flow cytometry |
Alexa Fluor 568 | 579 | 603 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
PE-eFluor 610 | 565 | 606 | Protein-based tandem dye | flow cytometry |
Texas Red (and Texas Red-X) | 595 | 613 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Yellow 610 | 551 | 614 | Phiton-based DNA scaffold dye | flow cytometry |
Alexa Fluor 594 | 590 | 618 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Yellow 660 | 550 | 664 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Yellow 690 | 551 | 691 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Yellow 700 | 551 | 701 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Yellow 730 | 551 | 730 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Yellow 755 | 552 | 755 | Phiton-based DNA scaffold dye | flow cytometry |
PE-Cyanine7 | 569 | 780 | Protein-based tandem dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
NovaFluor Red 660 | 637 | 658 | Phiton-based DNA scaffold dye | flow cytometry |
Allophycocyanin (APC) | 651 | 660 | Protein-based fluorophore | flow cytometry |
Cy5 | 647 | 665 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
eFluor 660 | 651 | 668 | Small organic dye | flow cytometry |
Alexa Fluor 647 | 650 | 671 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Red 685 | 637 | 685 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Blue 690 | 672 | 688 | Phiton-based DNA scaffold dye | flow cytometry |
Alexa Fluor 660 | 663 | 691 | Small organic dye | flow cytometry |
NovaFluor Red 700 | 637 | 701 | Phiton-based DNA scaffold dye | flow cytometry |
Alexa Fluor 680 | 681 | 704 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Red 710 | 637 | 713 | Phiton-based DNA scaffold dye | flow cytometry |
Alexa Fluor 700 | 696 | 719 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Red 725 | 636 | 725 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Red 755 | 636 | 755 | Phiton-based DNA scaffold dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
Alexa Fluor 750 | 752 | 776 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
APC-eFluor 780 | 756 | 785 | Protein-based tandem dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
Brilliant Ultra Violet 395 | 347 | 399 | Polymer dye | flow cytometry |
Alexa Fluor 350 | 343 | 441 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Brilliant Ultra Violet 496 | 348 | 493 | Tandem polymer dye | flow cytometry |
Qdot 525 Probe | 300 | 525 | Quantum dot | microscopy; flow cytometry |
Brilliant Ultra Violet 563 | 349 | 563 | Tandem polymer dye | flow cytometry |
Qdot 565 Probe | 300 | 564 | Quantum dot | microscopy; flow cytometry |
Qdot 605 Probe | 300 | 603 | Quantum dot | microscopy; flow cytometry |
Brilliant Ultra Violet 615 | 350 | 607 | Tandem polymer dye | flow cytometry |
Qdot 655 Probe | 300 | 654 | Quantum dot | microscopy; flow cytometry |
Brilliant Ultra Violet 661 | 349 | 659 | Tandem polymer dye | flow cytometry |
Qdot 705 Probe | 300 | 702 | Quantum dot | microscopy; flow cytometry |
Brilliant Ultra Violet 737 | 350 | 740 | Tandem polymer dye | flow cytometry |
Qdot 800 Probe | 300 | 792 | Quantum dot | microscopy; flow cytometry |
Brilliant Ultra Violet 805 | 349 | 804 | Tandem polymer dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
Alexa Fluor 405 | 401 | 422 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Brilliant Violet 421 | 406 | 423 | Polymer dye | flow cytometry |
Super Bright 436 | 413 | 431 | Polymer dye | flow cytometry |
eFluor 450 | 405 | 445 | Small organic dye | flow cytometry |
Pacific Blue | 404 | 455 | Small organic dye | flow cytometry |
Coumarin and Coumarin Derivatives | 387 | 470 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Brilliant Violet 480 | 437 | 478 | Polymer dye | flow cytometry |
Cyan Fluorescent Protein (CFP) | 435 | 485 | Expressed fluorescent protein | microscopy; flow cytometry |
eFluor 506 | 419 | 508 | Small organic dye | flow cytometry |
Pacific Green | 411 | 510 | Small organic dye | flow cytometry |
Pacific Orange | 400 | 551 | Small organic dye | flow cytometry |
Super Bright 600 | 414 | 601 | Tandem polymer dye | flow cytometry |
Brilliant Violet 605 | 405 | 605 | Tandem polymer dye | flow cytometry |
Super Bright 645 | 414 | 645 | Tandem polymer dye | flow cytometry |
Brilliant Violet 650 | 407 | 646 | Tandem polymer dye | flow cytometry |
Super Bright 702 | 413 | 702 | Tandem polymer dye | flow cytometry |
Brilliant Violet 711 | 406 | 711 | Tandem polymer dye | flow cytometry |
Super Bright 780 | 413 | 780 | Tandem polymer dye | flow cytometry |
Brilliant Violet 786 | 406 | 788 | Tandem polymer dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
Green Fluorescent Protein (GFP) | 488 | 510 | Expressed fluorescent protein | microscopy; flow cytometry |
BODIPY FL | 502 | 511 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Blue 510 | 493 | 513 | Phiton-based DNA scaffold dye | flow cytometry |
Fluorescein (FITC) | 498 | 517 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Alexa Fluor 488 | 499 | 520 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Oregon Green 488 | 498 | 526 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Blue 530 | 508 | 527 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Blue 555 | 493 | 555 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Blue 585 | 493 | 586 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Blue 610-30S | 508 | 615 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Blue 610-70S | 492 | 616 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Blue 660-40S | 508 | 664 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Blue 660-120S | 492 | 665 | Phiton-based DNA scaffold dye | flow cytometry |
PerCP-Cyanine5.5 | 489 | 679 | Protein-based tandem dye | flow cytometry |
PerCP-eFluor 710 | 482 | 708 | Protein-based tandem dye | flow cytometry |
NovaFluor Blue 725 | 492 | 725 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Blue 760 | 490 | 764 | Phiton-based DNA scaffold dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
Alexa Fluor 532 | 534 | 553 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Cy3 | 554 | 566 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Yellow 570 | 551 | 566 | Phiton-based DNA scaffold dye | flow cytometry |
Alexa Fluor 555 | 553 | 568 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Alexa Fluor 561 | 556 | 569 | Small organic dye | flow cytometry |
Alexa Fluor 546 | 561 | 572 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
R-phycoerythrin (R-PE) | 565 | 576 | Protein-based fluorophore | flow cytometry |
Tetramethylrhodamine (TRITC) | 552 | 578 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Red Fluorescent Protein (RFP) | 555 | 584 | Expressed fluorescent protein | microscopy; flow cytometry |
NovaFluor Yellow 590 | 563 | 594 | Phiton-based DNA scaffold dye | flow cytometry |
Alexa Fluor 568 | 579 | 603 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
PE-eFluor 610 | 565 | 606 | Protein-based tandem dye | flow cytometry |
Texas Red (and Texas Red-X) | 595 | 613 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Yellow 610 | 551 | 614 | Phiton-based DNA scaffold dye | flow cytometry |
Alexa Fluor 594 | 590 | 618 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Yellow 660 | 550 | 664 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Yellow 690 | 551 | 691 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Yellow 700 | 551 | 701 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Yellow 730 | 551 | 730 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Yellow 755 | 552 | 755 | Phiton-based DNA scaffold dye | flow cytometry |
PE-Cyanine7 | 569 | 780 | Protein-based tandem dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
NovaFluor Red 660 | 637 | 658 | Phiton-based DNA scaffold dye | flow cytometry |
Allophycocyanin (APC) | 651 | 660 | Protein-based fluorophore | flow cytometry |
Cy5 | 647 | 665 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
eFluor 660 | 651 | 668 | Small organic dye | flow cytometry |
Alexa Fluor 647 | 650 | 671 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Red 685 | 637 | 685 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Blue 690 | 672 | 688 | Phiton-based DNA scaffold dye | flow cytometry |
Alexa Fluor 660 | 663 | 691 | Small organic dye | flow cytometry |
NovaFluor Red 700 | 637 | 701 | Phiton-based DNA scaffold dye | flow cytometry |
Alexa Fluor 680 | 681 | 704 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Red 710 | 637 | 713 | Phiton-based DNA scaffold dye | flow cytometry |
Alexa Fluor 700 | 696 | 719 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Red 725 | 636 | 725 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Red 755 | 636 | 755 | Phiton-based DNA scaffold dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
Alexa Fluor 750 | 752 | 776 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
APC-eFluor 780 | 756 | 785 | Protein-based tandem dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
Brilliant Ultra Violet 395 | 347 | 399 | Polymer dye | flow cytometry |
Alexa Fluor 405 | 401 | 422 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Brilliant Violet 421 | 406 | 423 | Polymer dye | flow cytometry |
Super Bright 436 | 413 | 431 | Polymer dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
Alexa Fluor 350 | 343 | 441 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
eFluor 450 | 405 | 445 | Small organic dye | flow cytometry |
Pacific Blue | 404 | 455 | Small organic dye | flow cytometry |
Coumarin and Coumarin Derivatives | 387 | 470 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Brilliant Violet 480 | 437 | 478 | Polymer dye | flow cytometry |
Cyan Fluorescent Protein (CFP) | 435 | 485 | Expressed fluorescent protein | microscopy; flow cytometry |
Brilliant Ultra Violet 496 | 348 | 493 | Tandem polymer dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
eFluor 506 | 419 | 508 | Small organic dye | flow cytometry |
Green Fluorescent Protein (GFP) | 488 | 510 | Expressed fluorescent protein | microscopy; flow cytometry |
Pacific Green | 411 | 510 | Small organic dye | flow cytometry |
BODIPY FL | 502 | 511 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Blue 510 | 493 | 513 | Phiton-based DNA scaffold dye | flow cytometry |
Fluorescein (FITC) | 498 | 517 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Alexa Fluor 488 | 499 | 520 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Qdot 525 Probe | 300 | 525 | Quantum dot | microscopy; flow cytometry |
Oregon Green 488 | 498 | 526 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Blue 530 | 508 | 527 | Phiton-based DNA scaffold dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
Pacific Orange | 400 | 551 | Small organic dye | flow cytometry |
Alexa Fluor 532 | 534 | 553 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Blue 555 | 493 | 555 | Phiton-based DNA scaffold dye | flow cytometry |
Brilliant Ultra Violet 563 | 349 | 563 | Tandem polymer dye | flow cytometry |
Qdot 565 Probe | 300 | 564 | Quantum dot | microscopy; flow cytometry |
Cy3 | 554 | 566 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Yellow 570 | 551 | 566 | Phiton-based DNA scaffold dye | flow cytometry |
Alexa Fluor 555 | 553 | 568 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Alexa Fluor 561 | 556 | 569 | Small organic dye | flow cytometry |
Alexa Fluor 546 | 561 | 572 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
R-phycoerythrin (R-PE) | 565 | 576 | Protein-based fluorophore | flow cytometry |
Tetramethylrhodamine (TRITC) | 552 | 578 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Red Fluorescent Protein (RFP) | 555 | 584 | Expressed fluorescent protein | microscopy; flow cytometry |
NovaFluor Blue 585 | 493 | 586 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Yellow 590 | 563 | 594 | Phiton-based DNA scaffold dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
Super Bright 600 | 414 | 601 | Tandem polymer dye | flow cytometry |
Alexa Fluor 568 | 579 | 603 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Qdot 605 Probe | 300 | 603 | Quantum dot | microscopy; flow cytometry |
Brilliant Violet 605 | 405 | 605 | Tandem polymer dye | flow cytometry |
PE-eFluor 610 | 565 | 606 | Protein-based tandem dye | flow cytometry |
Brilliant Ultra Violet 615 | 350 | 607 | Tandem polymer dye | flow cytometry |
Texas Red (and Texas Red-X) | 595 | 613 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Yellow 610 | 551 | 614 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Blue 610-30S | 508 | 615 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Blue 610-70S | 492 | 616 | Phiton-based DNA scaffold dye | flow cytometry |
Alexa Fluor 594 | 590 | 618 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
Super Bright 645 | 414 | 645 | Tandem polymer dye | flow cytometry |
Brilliant Violet 650 | 407 | 646 | Tandem polymer dye | flow cytometry |
Qdot 655 Probe | 300 | 654 | Quantum dot | microscopy; flow cytometry |
NovaFluor Red 660 | 637 | 658 | Phiton-based DNA scaffold dye | flow cytometry |
Brilliant Ultra Violet 661 | 349 | 659 | Tandem polymer dye | flow cytometry |
Allophycocyanin (APC) | 651 | 660 | Protein-based fluorophore | flow cytometry |
NovaFluor Blue 660-40S | 508 | 664 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Yellow 660 | 550 | 664 | Phiton-based DNA scaffold dye | flow cytometry |
Cy5 | 647 | 665 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Blue 660-120S | 492 | 665 | Phiton-based DNA scaffold dye | flow cytometry |
eFluor 660 | 651 | 668 | Small organic dye | flow cytometry |
Alexa Fluor 647 | 650 | 671 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
PerCP-Cyanine5.5 | 489 | 679 | Protein-based tandem dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
NovaFluor Red 685 | 637 | 685 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Blue 690 | 672 | 688 | Phiton-based DNA scaffold dye | flow cytometry |
Alexa Fluor 660 | 663 | 691 | Small organic dye | flow cytometry |
NovaFluor Yellow 690 | 551 | 691 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Red 700 | 637 | 701 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Yellow 700 | 551 | 701 | Phiton-based DNA scaffold dye | flow cytometry |
Qdot 705 Probe | 300 | 702 | Quantum dot | microscopy; flow cytometry |
Super Bright 702 | 413 | 702 | Tandem polymer dye | flow cytometry |
Alexa Fluor 680 | 681 | 704 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
PerCP-eFluor 710 | 482 | 708 | Protein-based tandem dye | flow cytometry |
Brilliant Violet 711 | 406 | 711 | Tandem polymer dye | flow cytometry |
NovaFluor Red 710 | 637 | 713 | Phiton-based DNA scaffold dye | flow cytometry |
Alexa Fluor 700 | 696 | 719 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Red 725 | 636 | 725 | Phiton-based DNA scaffold dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
NovaFluor Yellow 730 | 551 | 730 | Phiton-based DNA scaffold dye | flow cytometry |
Brilliant Ultra Violet 737 | 350 | 740 | Tandem polymer dye | flow cytometry |
NovaFluor Red 755 | 636 | 755 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Yellow 755 | 551 | 755 | Phiton-based DNA scaffold dye | flow cytometry |
Alexa Fluor 750 | 752 | 776 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
PE-Cyanine7 | 569 | 780 | Protein-based tandem dye | flow cytometry |
Super Bright 780 | 413 | 780 | Tandem polymer dye | flow cytometry |
APC-eFluor 780 | 756 | 785 | Protein-based tandem dye | flow cytometry |
Brilliant Violet 786 | 406 | 788 | Tandem polymer dye | flow cytometry |
Qdot 800 Probe | 300 | 792 | Quantum dot | microscopy; flow cytometry |
Brilliant Ultra Violet 805 | 349 | 804 | Tandem polymer dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
Brilliant Ultra Violet 395 | 347 | 399 | Polymer dye | flow cytometry |
Alexa Fluor 405 | 401 | 422 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Brilliant Violet 421 | 406 | 423 | Polymer dye | flow cytometry |
Super Bright 436 | 413 | 431 | Polymer dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
Alexa Fluor 350 | 343 | 441 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
eFluor 450 | 405 | 445 | Small organic dye | flow cytometry |
Pacific Blue | 404 | 455 | Small organic dye | flow cytometry |
Coumarin and Coumarin Derivatives | 387 | 470 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Brilliant Violet 480 | 437 | 478 | Polymer dye | flow cytometry |
Cyan Fluorescent Protein (CFP) | 435 | 485 | Expressed fluorescent protein | microscopy; flow cytometry |
Brilliant Ultra Violet 496 | 348 | 493 | Tandem polymer dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
eFluor 506 | 419 | 508 | Small organic dye | flow cytometry |
Green Fluorescent Protein (GFP) | 488 | 510 | Expressed fluorescent protein | microscopy; flow cytometry |
Pacific Green | 411 | 510 | Small organic dye | flow cytometry |
BODIPY FL | 502 | 511 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Blue 510 | 493 | 513 | Phiton-based DNA scaffold dye | flow cytometry |
Fluorescein (FITC) | 498 | 517 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Alexa Fluor 488 | 499 | 520 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Qdot 525 Probe | 300 | 525 | Quantum dot | microscopy; flow cytometry |
Oregon Green 488 | 498 | 526 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Blue 530 | 508 | 527 | Phiton-based DNA scaffold dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
Pacific Orange | 400 | 551 | Small organic dye | flow cytometry |
Alexa Fluor 532 | 534 | 553 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Blue 555 | 493 | 555 | Phiton-based DNA scaffold dye | flow cytometry |
Brilliant Ultra Violet 563 | 349 | 563 | Tandem polymer dye | flow cytometry |
Qdot 565 Probe | 300 | 564 | Quantum dot | microscopy; flow cytometry |
Cy3 | 554 | 566 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Yellow 570 | 551 | 566 | Phiton-based DNA scaffold dye | flow cytometry |
Alexa Fluor 555 | 553 | 568 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Alexa Fluor 561 | 556 | 569 | Small organic dye | flow cytometry |
Alexa Fluor 546 | 561 | 572 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
R-phycoerythrin (R-PE) | 565 | 576 | Protein-based fluorophore | flow cytometry |
Tetramethylrhodamine (TRITC) | 552 | 578 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Red Fluorescent Protein (RFP) | 555 | 584 | Expressed fluorescent protein | microscopy; flow cytometry |
NovaFluor Blue 585 | 493 | 586 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Yellow 590 | 563 | 594 | Phiton-based DNA scaffold dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
Super Bright 600 | 414 | 601 | Tandem polymer dye | flow cytometry |
Alexa Fluor 568 | 579 | 603 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Qdot 605 Probe | 300 | 603 | Quantum dot | microscopy; flow cytometry |
Brilliant Violet 605 | 405 | 605 | Tandem polymer dye | flow cytometry |
PE-eFluor 610 | 565 | 606 | Protein-based tandem dye | flow cytometry |
Brilliant Ultra Violet 615 | 350 | 607 | Tandem polymer dye | flow cytometry |
Texas Red (and Texas Red-X) | 595 | 613 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Yellow 610 | 551 | 614 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Blue 610-30S | 508 | 615 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Blue 610-70S | 492 | 616 | Phiton-based DNA scaffold dye | flow cytometry |
Alexa Fluor 594 | 590 | 618 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
Super Bright 645 | 414 | 645 | Tandem polymer dye | flow cytometry |
Brilliant Violet 650 | 407 | 646 | Tandem polymer dye | flow cytometry |
Qdot 655 Probe | 300 | 654 | Quantum dot | microscopy; flow cytometry |
NovaFluor Red 660 | 637 | 658 | Phiton-based DNA scaffold dye | flow cytometry |
Brilliant Ultra Violet 661 | 349 | 659 | Tandem polymer dye | flow cytometry |
Allophycocyanin (APC) | 651 | 660 | Protein-based fluorophore | flow cytometry |
NovaFluor Blue 660-40S | 508 | 664 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Yellow 660 | 550 | 664 | Phiton-based DNA scaffold dye | flow cytometry |
Cy5 | 647 | 665 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Blue 660-120S | 492 | 665 | Phiton-based DNA scaffold dye | flow cytometry |
eFluor 660 | 651 | 668 | Small organic dye | flow cytometry |
Alexa Fluor 647 | 650 | 671 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
PerCP-Cyanine5.5 | 489 | 679 | Protein-based tandem dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
NovaFluor Red 685 | 637 | 685 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Blue 690 | 672 | 688 | Phiton-based DNA scaffold dye | flow cytometry |
Alexa Fluor 660 | 663 | 691 | Small organic dye | flow cytometry |
NovaFluor Yellow 690 | 551 | 691 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Red 700 | 637 | 701 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Yellow 700 | 551 | 701 | Phiton-based DNA scaffold dye | flow cytometry |
Qdot 705 Probe | 300 | 702 | Quantum dot | microscopy; flow cytometry |
Super Bright 702 | 413 | 702 | Tandem polymer dye | flow cytometry |
Alexa Fluor 680 | 681 | 704 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
PerCP-eFluor 710 | 482 | 708 | Protein-based tandem dye | flow cytometry |
Brilliant Violet 711 | 406 | 711 | Tandem polymer dye | flow cytometry |
NovaFluor Red 710 | 637 | 713 | Phiton-based DNA scaffold dye | flow cytometry |
Alexa Fluor 700 | 696 | 719 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
NovaFluor Red 725 | 636 | 725 | Phiton-based DNA scaffold dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Fluorophore | Excitation | Emission | Chemical property | Platform |
---|---|---|---|---|
NovaFluor Yellow 730 | 551 | 730 | Phiton-based DNA scaffold dye | flow cytometry |
Brilliant Ultra Violet 737 | 350 | 740 | Tandem polymer dye | flow cytometry |
NovaFluor Red 755 | 636 | 755 | Phiton-based DNA scaffold dye | flow cytometry |
NovaFluor Yellow 755 | 551 | 755 | Phiton-based DNA scaffold dye | flow cytometry |
Alexa Fluor 750 | 752 | 776 | Small organic dye | microscopy; flow cytometry; fluorescence microplate reader |
PE-Cyanine7 | 569 | 780 | Protein-based tandem dye | flow cytometry |
Super Bright 780 | 413 | 780 | Tandem polymer dye | flow cytometry |
APC-eFluor 780 | 756 | 785 | Protein-based tandem dye | flow cytometry |
Brilliant Violet 786 | 406 | 788 | Tandem polymer dye | flow cytometry |
Qdot 800 Probe | 300 | 792 | Quantum dot | microscopy; flow cytometry |
Brilliant Ultra Violet 805 | 349 | 804 | Tandem polymer dye | flow cytometry |
Fluorescence SpectraViewer—compare the spectra and spectral properties of multiple fluorophores
Panel Builder—choose the optimal combination of fluorophores for your flow cytometry experiments
Stain-iT Cell Staining Simulator—visualize staining your cell without wasting your reagents, antibodies, or time
EVOS Light Cube Selection Guide—choose the light cube that best matches your dye
Alexa Fluor dyes are the leading and most trusted fluorescent dyes. These high-performing dyes have intense fluorescence with conjugates available across the visible spectrum.
Brilliant Ultra Violet (BUV) and Brilliant Violet (BV) dyes use polymer technology to offer a broad range of conjugated antibodies for full use of UV and violet detector channels.
eFluor conjugated antibodies were developed for superior performance in multiple applications, including multicolor flow cytometry and immunofluorescence microscopy.
NovaFluor dyes use the Phiton technology to create unique fluorescent signatures with minimal cross-excitation between laser lines resulting in lower need for compensation with increased resolution.
Super Bright dyes are comparable to Brilliant Violet dyes in terms of brightness while providing additional marker and clone options for violet-laser excitable antibody conjugates.
Qdot probes offer bright fluorescence performance for single-excitation multicolor analysis with long-term photostability for live-cell imaging, fixability for post in vivo studies, and narrow emission spectra for low crosstalk.
Protocols that fit your needs in flow cytometry ranging from sample preparation to numerous cell stimulation conditions, staining, immunophenotyping, and data analysis strategies.
A handy reference poster featuring the broad range of our dyes and labeling reagents available.
Protocols that fit your needs in imaging ranging from sample and assay preparation to staining, labeling, and data analysis strategies.
CyTM is a trademark or registered trademark of GE Healthcare.
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and is used under license.
For Research Use Only. Not for use in diagnostic procedures.