Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
The integrity of your science is important whether its in the lab or in transit. Smart-Tracker can help keep you aware of sample status in the lab and on the move.
Thermo Scientific understands the importance of maintaining product integrity, reducing loss and conserving money beyond your facility and created Smart-Tracker data loggers as a reliable and easy-to-use solution for temperature monitoring in equipment and throughout the cold chain.
Smart-Tracker data logger mission-based reporting provides detailed system and payload tracking, while the optional cloud application allows for remote monitoring, fleet analysis and documentation. Smart-Tracker data loggers allow you to program high and low temperature limits and can be an ideal solution for monitoring temperature in packages, refrigerated trucks, laboratories, storage facilities and more.
† Subject to network speed and availability
Simplified cold-chain assurance
Smart-Tracker monitors temperature in storage and transit, logging up to 4,000 readings and does not require dedicated computer equipment or complicated installation. Simply attach or insert the Smart-Tracker, no other installation required.
Smartphone application
Easy to use iOS and Android smartphone application enables intuitive, simple Smart-Tracker programming and remote monitoring.
Email alerts and notification
Smart Tracker provides datalogging mission status notifications and temperature alerts by email.
Bluetooth connectivity and cloud enablement
Smart-Trackers connect wirelessly via Bluetooth to your smart-phone or tablet with the Smart-Tracker app. From the app, you can start or stop datalogging missions, view data and email results. Optionally, upload the information including geo location provided by the smartphone to the Cloud, where authorized parties can check the latest status as little as a few seconds.
For Research Use Only. Not for use in diagnostic procedures.