Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Pipettes require routine servicing, compliance, and preventative maintenance throughout the year to ensure they perform optimally in line with industry standards. Ensuring pipettes are calibrated accurately and precisely is also an essential part of managing a lab’s assets, as calibrating your instruments can extend their product life cycle and reduce the total cost of ownership.
For accurate and precise pipette calibration that aligns with industry-specific standards, customers can rely on Thermo Fisher Scientific’s calibration services and support team and field service engineers (FSEs) for remote and onsite pipette calibration services that deliver first-class results. So, you can focus on what really matters – the science.
Whether a pipette check for cGLP, cGMP, academic research, or clinical testing, Thermo Fisher Scientific’s calibration services and support team are trained, and certified to provide calibration, repair and maintenance services for pipettes and pipetting instruments. We can calibrate, service and repair all makes and models of Thermo Scientific pipettes, as well as pipetting instruments from other suppliers.
Australia
Our pipette calibration service centre in Scoresby, Victoria is NATA accredited for all pipette calibrations from volumes 0.2 μL to 60 mL, providing customers with confidence that their valuable samples are being dispensed reliably– every time. All of our pipette services meet ISO 17025 and ISO 8655 compliance requirements, in line with Australian standards.
We offer the convenience of a single source for all your pipette service requirements:
|
|
|
New Zealand
Whether a pipette check for cGLP, cGMP, academic research, or clinical testing, Thermo Fisher Scientific’s calibration services and support team are trained, and certified to provide calibration, repair and maintenance services for pipettes and pipetting instruments. We can calibrate, service and repair all makes and models of Thermo Scientific pipettes, as well as pipetting instruments from other suppliers.
Please ensure you complete and attach the Pipette Service Request and Decontamination Declaration
when you send your pipettes to our service centres.
Finnpipettes are easy to service in the laboratory using the tools provided with the pipette. All Finnpipettes come with detailed instructions on how to disassemble and assemble the pipette. Instructions for routine in-lab maintenance of pipettes are also included. Instructions for pipette use are also available on our Pipetting Resources page.
At the beginning of each workday, the pipette should be checked for dust and dirt on the outside surfaces. Particular attention should be paid to the tip cone. No other solvents except 70% ethyl alcohol should be used to clean the pipette.
If the pipette is used daily, it is recommended to check the calibration every three months.
Please note: The calibration must be checked after cleaning the pipette.
All Finnpipette models are UV resistant. The handle however might change colour from grey to light yellow. If the inner parts of the pipette are exposed to UV light, please check there is sufficient grease on the piston and O-rings.