Search Thermo Fisher Scientific
Dye | Super Bright 436 | Super Bright 600 | Super Bright 645 | Super Bright 702 | Super Bright 780 |
---|---|---|---|---|---|
Comparable to | Brilliant Violet™ 421![]() | Brilliant Violet™ 605![]() | Brilliant Violet™ 650![]() | Brilliant Violet™ 711![]() | Brilliant Violet™ 786![]() |
Brightness |
|
|
|
|
|
Excitation | violet (405 nm) | violet (405 nm) | violet (405 nm) | violet (405 nm) | violet (405 nm) |
Emission | 436 nm | 600 nm | 645 nm | 702 nm | 780 nm |
Suggested bandpass filter | 450/50 | 610/20 | 660/20 | 710/50 | 780/60 |
More about the dye | View the Super Bright 436 dye dashboard | View the Super Bright 600 dye dashboard | View the Super Bright 645 dye dashboard | View the Super Bright 702 dye dashboard | View the Super Bright 780 dye dashboard |
Comparative data | See data | See data | See data | See data | See data |
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.