Blasticidin is a nucleoside antibiotic produced by the bacterium Streptomyces griseochromogenes. It is a potent translational inhibitor in both prokaryotic and eukaryotic cells. Resistance to blasticidin is conferred by the product of the bsd gene from Aspergillus terreus. Blasticidin has a fast, potent mode of action, causing rapid cell death at low antibiotic concentrations. E. coli strains are generally sensitive to concentrations of 50 µg/ml, while mammalian cells are sensitive to concentrations as low as 2 to 10 µg/ml. Cell death occurs rapidly, and blasticidin-resistant stable mammalian cell lines can be generated in less than one week.
Table 1 - Suggested Blasticidin Concentrations
Cell Line Concentration [ug/ml] Reference
HT1080
(human fibrosarcoma)
1-5
10
20
J Biol Chem283:11556–11564 (2008)
J Virol82:7325–7335 (2008)
Mol Biol Cell 19:8–16 (2008)
HeLa
(human cervical cancer)
10
10
20
J Gen Virol89:2611–2621 (2008)
Mol Biol Cell19:8–16 (2008).
J Immunol 181:22–26(2008)
HEK293
(human embryonic kidney)
5
10
J Biol Chem283:27534–27546 (2008)
J Virol82:1665–1678 (2008)
Hep G2
(human hepatocellular carcinoma)
4
5
J Biol Chem283:16320–16331 (2008)
Blood113: 1786–1793 (2009)
MCF-7
(human breast cancer)
2
5
Mol Cancer Res6:555–567 (2008)
Mol Endocrinol22: 361–379 (2008)
A549 (lung cancer)
10
Cancer Res68:5040–5048 (2008)