Search Thermo Fisher Scientific
Hygromycin B is an aminoglycosidic antibiotic that inhibits protein synthesis by disrupting translocation and promoting mistranslation at the 80S ribosome. Because it uses a different mode of action than Geneticin®, Blasticidin S, or Zeocin™, it’s perfect for dual-selection experiments when used in conjunction
with another selection agent.
Resistance to Hygromycin B is conferred by the E. coli hygromycin resistance gene (hyg or hph). The concentration for selection ranges from 100-1000 µg/ml (typically 200 µg/ml) and should be optimized for each cell line.
Formula: C20H37N3O13
FW: 527.5 g/mole
Cell types | Conc. (μg/ml)* | Invitrogen Vectors carrying resistance marker |
HeLa | ~550 | pcDNA5 vectors (Constitutive mammalian expression) |
CHO | ~250 | pIND/Hygro vector (Ecdysone-Inducible mammalian expression) |
Jurkat T-Cell | ~1,000 | pSecTag2/Hygro vector (Secreted mammalian expression) |
S2 Drosophila | 200-300 | pREP vectors (Episomal mammalian expression) |
pCoHygro (Selection vector for DES®) |
* The optimal concentration for selection of your cell line should be determined using kill curves.