Search Thermo Fisher Scientific
Cell Line | Concentration [µg/ml] | Reference |
---|---|---|
HT1080 (human fibrosarcoma) | 0.2–0.4 1 2 | J Biol Chem282:29314–29322 (2007) J Virol82:3320–3328 (2008) Cancer Res68: 1417–1426 (2008) |
HeLa (human cervical cancer) | 1-2 2 2 | Genes Cells12:397–406 (2007) Mol Cell Biol28:4104–4115 (2008) PNAS105:16490–16495 (2008) |
HEK293 (human embryonic kidney) | 0.25 2 3 | Nucleic Acids Res36:e83 (2008) Mol Cell Biol28:4104–4115 (2008) Mol Cell Biol 27:4708–4719 (2007) |
Hep G2 (human hepatocellular carcinoma) | 0.3 2 2 | J Biol Chem283:21462–21468 (2008) Mol Cell Biol28:4104–4115 (2008) J Biol Chem283:708–715 (2008) |
Human embryonic stem (ES) cells | 0.5 1 5 | Nucleic Acids Res 36:e148 (2008) Stem Cells26:850–863 (2008) Stem Cells26:2245–2256 (2008) |
MCF-7 (human breast cancer) | 0.45 1 2 | RNA13:1375-1383 (2007) Cancer Res68:1319–1328 (2008) Mol Cell Biol28:4104–4115 (2008) |
H1299 (non-small cell lung carcinoma) | 1 1.5 2.5 | Nucleic Acids Res 35:e17 (2007) J Biol Chem283:33394–33405 (2008) Proc Natl Acad Sci USA 105:1937–1942 (2008) |
MDA-MB-231 (human breast cancer) | 0.5 1.5 5 | Mol Cell Biol28:997–1006 (2008) Mol Cell Biol 27:324 – 339 (2007) Mol Cell Biol 28:687–704 (2008) |
A549 (lung cancer) | 1.5 1.5 | Mol Cell Biol 27:324–339 (2007) J Biol Chem283:33394–33405 (2008) |