Search Thermo Fisher Scientific
Commonly know as G418 or G-418, Geneticin® reagent is an aminoglycoside related to Gentamicin, and is commonly used as a selective agent for eukaryotic cells. As an analog of neomycin sulfate, it interferes with the function of 80S ribosomes and protein synthesis in eukaryotic cells.
The dominant-acting resistance genes (neor) used on common mammalian expression vectors contain elements derived from either transposons Tn601 (903) or Tn5. When grown in medium containing Geneticin® selection agent, stable colonies of mammalian cells expressing these resistance markers can be generated in 10 to 14 days.
To be effective, selection agents need to be reliable, highly pure, and consistent in nature. Geneticin® selective antibiotic, considered the gold standard for nearly two decades, is the only G-418 reagent that delivers these qualities to your mammalian cell cultures.
Compared to other manufacturers’ G-418 products, Geneticin® antibiotic exhibits higher purity, so you can use less and produce greater selection pressure with minimal toxicity from contaminants.
When evaluating G-418 products, consider more than just potency. Look at purity and working range. Taken together, these three characteristics provide a true evaluation of the effectiveness of your G-418.
Purity. The purity of Geneticin® selection agent is consistently greater than 90%, as determined by HPLC — far higher than any other supplier. This means:
ED50 Assay. This is a true measure of eukaryotic growth selectivity as determined on reference NIH3T3 cells. Higher purity generally translates to higher ED50 values.
This means when purchasing a new lot of Geneticin®, you do not need to re-optimize for ideal antibiotic concentration (assuming that no other media alterations). The wide ranges of ED50 displayed in different lots from alternative G-418 suppliers may require you to re-optimize for each lot. Lot-to-lot consistency with Geneticin® is sufficiently high that you will not notice any difference in selection performance between lots.
Purity. The purity of Geneticin® selection agent is consistently greater than 90%, as determined by HPLC — far higher than any other supplier. This means:
ED50 Assay. This is a true measure of eukaryotic growth selectivity as determined on reference NIH3T3 cells. Higher purity generally translates to higher ED50 values.
This means when purchasing a new lot of Geneticin®, you do not need to re-optimize for ideal antibiotic concentration (assuming that no other media alterations). The wide ranges of ED50 displayed in different lots from alternative G-418 suppliers may require you to re-optimize for each lot. Lot-to-lot consistency with Geneticin® is sufficiently high that you will not notice any difference in selection performance between lots.
Specification | Invitrogen | Supplier A | Supplier B | The Geneticin® Advantage |
Purity (HPLC) | >90-93% | 66-75% | 65-82% | Significantly higher purity |
Potency claimed (µg/mg) | 718-735 | 712-724 | 673-735 | More stringent potency assay |
Potency upon retest (µg/mg) | 718-735 | 640-659 | 621-677 | Consistently higher actual potency |
ED50 Assay (µg/ml) | 2450-2700 | 1350-3100 | 600-2350 | Reliable selection lot-to-lot |
Table 1. Multiple lots of G-418 from two different suppliers were analyzed according to Invitrogen raw material QC protocols for Geneticin®. It was observed that results obtained with G-418 from Supplier A and Supplier B displayed wide variations on all measured parameters, including an inconsistency between the actual potency and the value reported on the supplied product literature. Discrepancies between re-tested potency values and that claimed by other suppliers may be due to differences in G-418 reference lots and assay procedures.
Figure 1. Comparison of HPLC data of Geneticin® with selected lots of G-418 from Supplier A and Supplier B. Samples were prepared and analyzed according to the procedure published in Clinical Chem. (1978), Vol. 24, No.11, p1940.