Search Thermo Fisher Scientific
A thermal cycler (also called thermocycler) is an instrument that employs precise temperature control and rapid temperature changes to conduct the polymerase chain reaction (PCR). PCR is used to create billions of copies of a DNA sequence for use in a wide array of downstream procedures including cloning, sequencing, expression analysis, and genotyping. Our engineers have been designing and manufacturing high-quality Applied Biosystems® thermal cyclers since 1987; these instruments are renowned for their reliability, accuracy, and user-friendly interfaces.
Economical option for everyday cycling | Elegent design, smart choice | For robust thermal cycling and diagnostic applications | Most flexible and highest throughput | |
---|---|---|---|---|
Learn more | Learn more | Learn more | Learn more | |
2720 Thermal Cycler Demo request | NEW SimpliAmp™ Thermal Cycler Demo request | Veriti® Thermal Cycler** Demo request | ProFlex™ PCR System Demo request | |
Block formats |
|
|
|
|
Temperature optimization | None |
|
|
|
User inferface | Keypad | 8.4 in. touch screen | 6.5 in. touch screen | 8.4 in. touch screen |
Sample throughput | Up to 96 samples | Up to 96 samples | Up to 384 samples | Up to 480,000 samples |
Max block ramp rate*** | 2.3°C/sec | 4.0°C/sec | 5.0°C/sec | 6.0°C/sec |
**Also available as an FDA Class 1/ CE-IVD labeled device, click here for details
***For information about how ramp rates are calculated, click here
Looking for the GeneAmp® 9700 PCR System? Click here
Life Technologies provides award-winning service options for Applied Biosystems® Thermal Cyclers.
Support
Service plans