Search Thermo Fisher Scientific
Elegant design, smart choiceThe SimpliAmp™ Thermal Cycler is an easy-to-use, accurate, and small thermal cycler designed to fit any lab’s needs for everyday PCR. It is designed with reliable Applied Biosystems® engineering, including innovations like the responsive color touch screen and VeriFlex™ technology for simple and accurate optimization. |
|
|
|
|
|
|
VeriFlex™ Blocks provide a better-than-gradient approach to PCR optimization. With three separate Peltier blocks, you have precise control over three temperature zones.
Simple, intuitive touch screen and simulation modes
The SimpliAmp™ Thermal Cycler has a new, large color graphical user interface for easy programming and quick checks of your run status at the bench. Simulation modes now make the transition from previous thermal cyclers to the SimpliAmp™ Thermal Cycler simple, accurate, and efficient.
Innovation continues with the SimpliAmp™ Thermal Cycler, the newest Applied Biosystems® PCR instrument that allows you to interact with it from your desk, from the next lab, during a lab meeting, from your house, or wherever your schedule demands. Using your mobile device, you can check your run status and instrument availability, which helps make doing PCR with the SimpliAmp™ Thermal Cycler easy and efficient.
PCR Essentials app with built-in |
For Research Use Only. Not for use in diagnostic procedures. The information in this guide is subject to change without notice.