Search Thermo Fisher Scientific
Flexible configuration and controlThe ProFlex™ PCR System combines the reliability and performance you’ve come to expect from Applied Biosystems® thermal cyclers with the flexible configuration and control features that fit how you work today. Dual block configurations allow you to maximize your throughput. Request a demo |
The only thermal cycler that allows simultaneous remote access by three users to control three independent blocks, the ProFlex™ PCR System helps keep your lab’s PCR experiments visible, controllable, and on track.
|
| |
|
| |
|
Need Installation Qualification and Operation Qualification (IQ/OQ) help for your ProFlex™ PCR System? Click here to view our services.
The ProFlex™ PCR System has 5 different blocks that can be changed with the flip of a switch, including the first-of-its-kind 3 x 32-well block. This block allows up to three experiments to be run simultaneously, completely independently of each other and without interference. Dual 96-well and dual 384-well blocks are available for your high-throughput sequencing needs, and a dual flat block is available to support OpenArray® plate technology for genotyping analysis on the QuantStudio™ 12K Flex real-time PCR system.
3 x 32-well block | 96-well block | |
2 x 96-well block | 2 x 384-well block | |
2 x flat block |
Unparalleled innovation has been built into our new ProFlex™ PCR System, the only PCR instrument that allows you to interact with it from your desk, from the next lab, during a lab meeting, from your house, or wherever your schedule demands. Check your run status or block availability from your mobile device. Remote monitoring makes doing PCR easy and efficient.
PCR Essentials app with built-in My ProFlex™ remote: Download the app now > |
The ProFlex™ PCR System has a new, state-of-the-art graphical user interface for easy programming and quick checks of your run status at the bench. Simulation modes now make the transition from previous thermal cyclers to the ProFlex™ System simple, accurate, and efficient.
The ProFlex™ PCR System has 5 different blocks that can be changed with the flip of a switch, including the first-of-its-kind 3 x 32-well block. This block allows up to three experiments to be run simultaneously, completely independently of each other and without interference. Dual 96-well and dual 384-well blocks are available for your high-throughput sequencing needs, and a dual flat block is available to support OpenArray® plate technology for genotyping analysis on the QuantStudio™ 12K Flex real-time PCR system.
3 x 32-well block | 96-well block | |
2 x 96-well block | 2 x 384-well block | |
2 x flat block |
Unparalleled innovation has been built into our new ProFlex™ PCR System, the only PCR instrument that allows you to interact with it from your desk, from the next lab, during a lab meeting, from your house, or wherever your schedule demands. Check your run status or block availability from your mobile device. Remote monitoring makes doing PCR easy and efficient.
PCR Essentials app with built-in My ProFlex™ remote: Download the app now > |
The ProFlex™ PCR System has a new, state-of-the-art graphical user interface for easy programming and quick checks of your run status at the bench. Simulation modes now make the transition from previous thermal cyclers to the ProFlex™ System simple, accurate, and efficient.
Thermal cyclers: key thermal concepts and ramp rates
Thermal cycler comparison: the ProFlex™ 96-Well PCR System demonstrates consistent, reliable performance