Search Thermo Fisher Scientific
The Veriti® Thermal Cycler delivers the proven reliability you expect from Applied Biosystems® PCR instruments, plus the added control of VeriFlex™ Blocks. Veriflex™ Blocks offer you 6 independent temperature blocks providing precise control over your PCR optimization, supplying “better-than-gradient” functionality.
The powerful user interface, accessible via a color touch screen, simplifies the setup and use of the Veriti® 96-Well Fast Thermal Cycler. Optional setups for Fast or standard PCR methods provide you the flexibility to shorten your PCR cycling time.
VeriFlex™ Blocks provide a better-than-gradient approach to PCR optimization. With 6 separate Peltier blocks, you have precise control over 6 temperature zones.
Figure 1. Get linear gradient with Veriti Better than Gradient and not a non-linear curve.
Figure 2. (A) Screenshot from Veriti™ Thermal Cycler showing the annealing temperature being set for 6 different primers. (B) Six different annealing temperatures were set for each of the zones, with the annealing/extension step matching the length of the longest amplicon.
![]() | Control Veriti® Systems from Anywhere and EverywhereOptional VeritiLink™ Remote Management Software (VRMS) allows you to manage and monitor over 50 networked Veriti® Thermal Cyclers from your browser. With convenient features like real-time email updates, VRMS puts control at your fingertips. Learn more about VeritiLink™ Remote Management Software |
VeriFlex™ Blocks provide a better-than-gradient approach to PCR optimization. With 6 separate Peltier blocks, you have precise control over 6 temperature zones.
Figure 1. Get linear gradient with Veriti Better than Gradient and not a non-linear curve.
Figure 2. (A) Screenshot from Veriti™ Thermal Cycler showing the annealing temperature being set for 6 different primers. (B) Six different annealing temperatures were set for each of the zones, with the annealing/extension step matching the length of the longest amplicon.
![]() | Control Veriti® Systems from Anywhere and EverywhereOptional VeritiLink™ Remote Management Software (VRMS) allows you to manage and monitor over 50 networked Veriti® Thermal Cyclers from your browser. With convenient features like real-time email updates, VRMS puts control at your fingertips. Learn more about VeritiLink™ Remote Management Software |
Life Technologies provides award-winning service options for all Applied Biosystems® thermal cyclers.
Veriti® 96-Well Fast Thermal Cycler | Veriti® 96-Well Thermal Cycler | Veriti® 384-Well Thermal Cycler | Veriti® 60-Well Thermal Cycler | |
---|---|---|---|---|
Block Format | 0.1 mL Alloy | 0.2 mL Alloy | 0.02 mL Aluminum | 0.5 mL Aluminum |
Features | Fast 0.1 mL format and sample block provide enhanced performance and durability | Standard 0.2 mL format and sample block. Enabled to run FAST chemistry | Standard 0.02 mL format and sample block | Handles reaction tubes for large volume reactions |
Max Block Ramp Rate | 5.00 ºC/Sec | 3.9 ºC/Sec | 3.7º C/Sec | 3.3º C/Sec |
Max Sample Ramp Rate* | 4.25 ºC/Sec | 3.35 ºC/Sec | 3.1º C/Sec | 2.7º C/Sec |
Enabled to run FAST Chemistry | Yes | Yes | No | No |
Temperature Accuracy | ±0.25 ºC (35 ºC – 99.9 ºC) | |||
Temperature Range | 0ºC to 100.0ºC | |||
Temperature Uniformity | <0.5 ºC (20 sec after reaching 95 ºC) | |||
Dimensions | Height: 24.5cm (9.6 in.) Width: 23.7 cm (9.3 in.) Depth: 48.5 cm (19.1 in.) | |||
Weight | 11.4 kg (25 lb) | |||
PCR Volume Range | 10-30 µL | 10-80 µL | 5-20 μL | 25-100 μL |
Instrument Memory | USB and On-board | |||
Display Interface | 6.5” VGA 32k color with Touch Screen | |||
Tm Calculator | Menu driven through touch screen | |||
Power | 100-240V, 50-60Hz Max: 800 VA | |||
VeriFlex™ Blocks | 25 °C (5 °C Zone-to-Zone) | 25 °C (5 °C Zone-to-Zone) | N/A | N/A |
*For information about how Ramp rates are calculated, click here
Discover new Veriti® Thermal Cycler protocols with firmware upgrade