Search Thermo Fisher Scientific
通常情况下,我们通过加氯消毒、臭氧处理或氯胺消毒法对饮用水进行消毒处理,从而杀灭饮用水中的细菌和其他有机物。然而,消毒过程中会产生对人体健康有害的危险副产品,通常称为卤乙酸(HAAs)。EPA指出,若饮用水中所含的HAA水平超过0.06 mg/L(60 µg/L),则患癌的风险就会增加。在对HAAs进行频繁监测时,出现一种全新的方法(IC-MS/MS),该方法对于检测HAAs具有较高的灵敏度,同时其检出限低于以往EPA方法中的规定。