Search Thermo Fisher Scientific
Our portfolio of reusable support containers accommodates a range of functionalities and chamber dimensions. The interaction between the BPC and the outer container system is important in simple liquid-handling applications, where the BPC system is not shipped, and becomes even more critical when transportation takes place. We offer plastic and stainless steel options to suit your storage and transport needs.
Contain and store bioproduction fluids in these durable, production-ready, all-purpose vessels. Nalgene production tanks are economical alternatives to stainless-steel tanks. Available in a variety of sizes and shapes, ranging from 75 L to 380 L.
Rigid plastic vessels to support our BPC products in helping meet your large-volume liquid storage, handling, and shipping needs.
Durable outer support containers used for in-house large volume liquid handling operations. These containers are designed to allow both top and bottom discharge from BPCs.
Designed specifically for the storage and transport of active pharmaceutical ingredients and bulk intermediates and are available in PC, PETG, and HDPE, and in volumes ranging from 30 mL to 20 L.
When using carboys for critical applications, supporting documentation, choices and designs sensitive to the necessary cleaning validations are a few things to look for.
Standard and custom BPCs, tubing sets, and manifolds, covering a range of applications, sizes, and connection systems.
Our portfolio of reusable support containers accommodates a range of functionalities and chamber dimensions. The interaction between the BPC and the outer container system is important in simple liquid-handling applications, where the BPC system is not shipped, and becomes even more critical when transportation takes place. We offer plastic and stainless steel options to suit your storage and transport needs.
Useful for internal and transportation applications. Top drain only and top- and bottom-drain, high-density polyethylene cylindrical drums from 50 L to 300 L, as well as square or rectangular polypropylene containers from 200 to 1,000 L are available. Regular and cylindrical tanks are also available in HDPE, LLPDE and PP in sizes ranging from 75 L to 757 L.
Thermo Scientific HyPerforma Smartainer systems, with a range from 200 to 2,000 L. These containers are used for in-house, large-volume liquid-handling and storage operations.
Meet the challenges of your most demanding applications with formats, formulations, films, BPCs, and packaging designed to support your needs.
Meet the challenges of your most demanding applications with formats, formulations, films, BPCs, and packaging designed to support your needs.
点击以下链接获取产品手册、培训视频、手机App和更多资源。