Search Thermo Fisher Scientific
Hydrophobic interaction chromatography (HIC) is a versatile method for the purification and separation of biomolecules by using the function of hydrophobicity. The proteins containing both hydrophilic and hydrophobic regions are applied to a HIC column under specified salt buffer conditions, which promotes the binding of the biomolecule to the HIC resin but also has a stabilizing influence on the molecule’s structure. HIC steps are commonly used at all steps in the process including capture step, intermediate and final polish purification.
Register to watch this webinar about a process development case study using POROS HIC in bind-elute mode for high-aggregate mAb polishing. We will highlight the differences and specifics of the POROS HIC resins that address current purification challenges as more diversified classes of biomolecules being developed.
Thermo Fisher Scientific provides a suite of POROS HIC resins offering a differentiating range in hydrophobicity suitable for bind/elute and flow-through applications. The resins can be utilized at lower salt concentrations for the purification of a wide variety of biomolecules including therapeutic proteins, antibody fragments, antibody drug conjugates (ADCs), and other large molecules.
POROS HIC Resin | Surface Chemistry | Key Application | |
POROS Ethyl | Novel ethyl | Bind/elute mode of moderately to considerably hydrophobic molecules | |
POROS Benzyl | Low density benzyl/aromatic | Bind/elute or flow-through mode depending on molecule | |
POROS Benzyl Ultra | High density benzyl/aromatic | Flow-through mode in lower salt to bind impurities such as aggregates |
Figure 1: Order of hydrophobicity of POROS HIC resins. Hydrophobicity based on lysozyme gradient elution.
The new 50 µm resin backbone consists of cross‐linked poly(styrene-divinylbenzene) with a unique pore structure that provides rapid mass transport and enables enhanced productivity. The particle surface is coated with a novel polymer coating, which is then further derivatized with a range of hydrophobic ligands for flexible purification process design (Figure 2). POROS HIC resins deliver superior performance independent of flow rate, a major improvement in the purification tool box for impurity removal.
Figure 2. Separation of a mixture of standard proteins using POROS Ethyl, POROS Benzyl and POROS Benzyl Ultra resins. Column size: 0.46cmD X 20cmL; Protein Mixture: Ribonuclease A, Lysozyme, Chymotrypsin and Chymotrypsinogen in 1.7M ammonium sulfate, 50mM sodium phosphate pH 7.0; Gradient Elution: 1.7M ammonium sulfate/50 mM sodium phosphate pH 7.0 to 50 mM sodium phosphate pH 7.0 over 10 column volumes (CV); Flow rate: 100 cm/hr.
点击以下链接获取产品手册、培训视频、手机App和更多资源。
Pharmaceutical Grade Reagent. For Manufacturing and Laboratory Use Only.
For research use only. Not for use in diagnostic procedures.