Search Thermo Fisher Scientific
- 08/09/16
Antibodies are used in a broad range of research and diagnostic applications for the enrichment, detection, and quantitation of proteins and their modifications. Hundreds of thousands of antibodies are commercially available against thousands of proteins, which are used in a variety of applications, including western blotting (WB), immunofluorescence (IF), immunoprecipitation (IP), flow cytometry (FC), chromatin IP (ChIP), and enzyme-linked immunoassays (ELISA). These antibodies may be monoclonal, polyclonal, or recombinant from different organisms, and they may be used to interrogate biological systems and signaling pathways, diagnose disease, and assess responses to treatment [1].
Unfortunately, many antibodies are poorly characterized, both initially and between manufacturing lots. This is due to three challenges: 1) the sheer number of available protein targets and antibodies can be overwhelming to consider validating, so efficient and thorough characterization strategies are required; 2) the complex nature of proteins, binding interactions, physiochemical conditions (such as native versus denatured epitope conformations), and biology for each antibody application and model system, and; 3) the lack of consistent standard approaches and criteria to assess antibody selectivity [2, 3]. There is a great need to verify that antibodies recognize their intended targets and to ensure that the reagents are fit-for-purpose in a given application. A variety of antibody validation criteria have been proposed, including: 1) the verification of antibody specificity with genetic knockdowns or blocking peptides; 2) validation of antibody detection results with different biological systems (e.g., target localization, expression in model systems, etc.); 3) correlation of antibody results between methods, and; 4) demonstration of reproducibility between samples, labs, and manufacturing lots [3–5].
Recently, mass spectrometric (MS) approaches have been proposed for antibody target verification [6, 7]. Despite the cost and technical requirements of MS, of all existing validation methods, mass spectrometry has the unique ability to identify the actual antibody target(s), isoforms, post-translational modifications, and target-associated proteins present within a sample. Only MS can identify and characterize antibodies with this level of depth and specificity. Unlike western blotting, ELISA, and other standard immunological methods that use blocking reagents (such as milk or bovine serum albumin, BSA) to minimize background protein binding, mass spectrometry detects all proteins, specific and non-specific, in a prepared sample. For example, immunoprecipitation with an immobilized antibody on a bead or resin is a common approach to enrich a target protein and associated proteins from a lysate or biofluid. Due to the low abundance of a specific target relative to common background proteins, non-specifically bound proteins may overwhelm and interfere with or prevent the detection of a low abundance target. Even with stringent wash conditions and optimized reagents, dozens to hundreds of background proteins are typically observed in an immunoprecipitated sample that is analyzed by mass spectrometry. Thus, while MS holds great promise for contextualizing targets amidst potential interacting proteins, the MS results from an immunoprecipitated sample can be difficult to filter and interpret since non-specific background proteins will also be present. In particular, an assurance of antibody selectivity for a native protein from a real biological system is particularly challenging to demonstrate.
To address these issues and to assess the binding specificity of Invitrogen antibodies produced by Thermo Fisher Scientific, here we describe a new approach to antibody target verification. The benefits of this IP-MS approach include: identification of the antibody target(s), isoforms, and modifications, quantitative assessment of antibody selectivity by calculating fold-enrichment of targets and off-targets for the assessment of antibody selectivity, and identification of interacting proteins.
Thermo Fisher Scientific currently offers more than 48,000 antibodies to more than 20,000 proteins and protein modifications. The TP53 gene is the most highly referenced gene/protein in PubMed, with more than 7,500 references. TP53 is a key signaling protein with many known modifications and interacting proteins, and gain- and loss-of-function mutations in the TP53 gene are involved in genetic instability and tumorigenesis. The next 999 most- referenced genes/proteins vary greatly in their reference frequency, so the distribution of antibody protein targets has a very long “tail.” Antibody targets were chosen based on these considerations and criteria, and systematically verified through our IP-MS workflow.
図2. Representative list of the top 1,000 most-referenced genes in PubMed as of January 2016.
To identify candidate complementary cell lines for our selected protein targets, we utilized literature resources, public transcriptomic/proteomic databases, and biological samples likely to contain the most diverse and/or comprehensive set of protein targets to our prioritized pathways and panels. A combination of these resources was used to select 12 complementary cell lines likely to express more than 90% of the top 1,000 most-referenced genes or gene products for deep proteome analysis and IP-MS antibody verification.
図3. Selection of cell lines for antibody verification by IP-MS. A. RNA expression Z-scores from the NCI60 cell line panel were hierarchically clustered for 22 genes in the Ion Ampliseq Colon and Lung Cancer Panel. B. Venn diagram of the number of proteins identified from five NCI60 cell lines with mass spectrometric analysis of fractionated peptides from each lysate.
These 12 selected human cell lines were grown in recommended conditions in order to generate protein lysates from each cell line for MS-based proteome analysis. Using optimized sample preparation methods and instrumentation, we generated proteome data of unfractionated and fractionated samples to confirm that the cell lines expressed our selected target proteins. Briefly, lysate proteins were solubilized, proteolytically digested with trypsin, and prepared for LC-MS analysis. Digested protein samples were directly analyzed by LC-MS (unfractionated) as well as fractionated using high pH reverse phase columns to improve the depth of proteome coverage in subsequent LC-MS analyses. Each unfractionated protein digest identified 3,800–4,500 unique protein groups, and each fractionated protein digest identified 7,500–9,000 unique protein groups. To assess our overall protein sequence coverage and overlap between cell lines, the identified proteins from each cell line were then compared to determine pair-wise correlation scores. These correlations ranged from 75–95%.
To rapidly and effectively screen IP samples for the presence or absence of target proteins, we used the MS peptide peak areas of the top 3 peptides observed per protein, the number of unique peptides identified, and the spectral count detected with Thermo Scientific Proteome Discoverer Software (Cat. Using these metrics, target protein expression was ranked and compared across the 12 cell lines. IP-MS antibody verification used this protein expression data to select cell lines which expressed target proteins at medium to low abundance. CDH1 was detected only in unfractionated HCT116, LNCaP, and MCF7 cells, while CDH2 was only seen in unfractionated A549, BT549, HEK293, and Hs578T cells. Furthermore, cell lines with high expression of protein targets were deemed inappropriate models for antibody testing. Target proteins were ranked by their protein LFQ values in order to visualize protein target abundance within the context of the whole proteome and select appropriate cells lines for antibody screening. This protein expression information was invaluable for the selection of cell models and assessment of isoform-specific and pan-specific antibody selectivity. As a result, one or more cell lines for each target protein were chosen for this study based upon MS identification and target protein abundance.
図 4. Cell line selection for CDH1 and CDH2. A. Comparison of E-cadherin (CDH1) and N-cadherin (CDH2) protein expression across twelve cell lines without or with fractionation for deeper proteome analysis. B. Distribution of expressed proteins detected in unfractionated and fractionated MCF7 and A549 cells, highlighting the expression levels of CDH1 and CHD2.
We utilized protein expression profiles to assist in the assessment of antibodies by IP-MS. The key benefit of verifying an antibody’s target by IP-MS is identification of not only the native target protein, but also its isoforms, post-translational modifications, and interacting proteins. Historically, the results of this target identification can be assessed in several ways, including the number of unique peptides, protein sequence coverage, number of spectra observed for peptides from the target protein (spectral count), or integrated MS signal intensities from a subset or all of the detected peptides, as described above. The relative performance of various antibodies for the same target can be easily compared regardless of the measurement approach. Results of replicate IP samples were highly reproducible, and all 10 antibodies showed reproducible MS signals (CV<25% across replicates). This IP-MS approach assesses antibody fit-for-purpose, provides definitive evidence of target protein capture, and readily permits antibody comparisons that may indicate relative antibody affinity.
As mentioned previously, protein IP with immobilized antibodies is a common method for targeted protein enrichment, but tens to hundreds of background proteins are commonly identified by mass spectrometry even after stringent washing conditions. Quantitative tools to analyze protein affinity capture results, such as COMPASS, SAINT, and Perseus, offer sophisticated scoring methods and data visualization for filtering protein identifications, but the implementation of these methods can be challenging and the results difficult to interpret [7, 9–11]. To better understand these background proteins and more easily identify specifically captured versus non-specific proteins, we attempted to simplify the data representation by using protein LFQ values to quantitatively compare the proteins immunoprecipitated with a specific antibody versus a negative control antibody (Figure 6A). The resulting scatter plot of MS intensities had three clusters: This approach could be easily adapted to compare an antibody of interest to multiple negative control antibodies to remove common, non-specifically bound proteins. Interestingly, some of the common background proteins along the diagonal were significantly enriched with both antibodies. These could be due to specific binding to the magnetic bead resin or antibody isotype, and may depend on the cell type used in the sample preparation. This scatter plot approach typically eliminated more than 90% of the identified proteins as non-specific binders.
While the IP-MS approach provides verification of intended protein targets, it also identifies all other proteins present in an IP sample. This includes the IP antibody and carrier proteins, like BSA, in addition to interacting proteins and abundant non-specific proteins. These additional proteins present a challenge to verifying antibody selectivity and performance. To better quantify the performance and selectivity of an antibody and to normalize the results of antibodies against different targets across experiments and cell models, we utilized the concept of protein fold-enrichment. Calculations of fold-enrichment are commonly used to assess and optimize protein purification methods, and this approach can be used to assess an antibody’s ability to enrich its native target and interaction partners relative to background proteins from a biological matrix. The formula we used for this analysis is:
Cell lines were chosen for antibody assessment based upon a deep, MS-based proteome analysis, and more than 10,000 protein groups were identified and quantified in whole cell lysates. Using this data, antibody performance could be assessed quantitatively by calculating the fold-enrichment of all proteins identified in an IP sample. In this manner, the antibody target could be verified and the performance of different antibodies to the same target could be compared.
The pan-specific anti-cadherin antibody enriched R-cadherin (CDH4), E-cadherin (CDH1), and N-Cadherin (CDH2) by 30- to 80-fold. Interestingly, the TRIM9 protein was also enriched 50-fold, suggesting potential cross-reactivity with TRIM9. A previous bioinformatic analysis of TRIM9 and its related proteins highlighted regions of structural similarity to the cadherin superfamily of proteins, potentially explaining the capture of TRIM9 protein with this pan-specific antibody [13].
All six previously validated antibodies successfully captured the target, and an additional 8 antibodies not previously validated for IP also captured the target. APC promotes rapid degradation of CTNNB1, and both proteins play a key role in colorectal cancer [14]. The beta-catenin antibodies were screened with HCT116 cell lysate based upon the MS-based proteome expression profile of beta-catenin, and the HCT116 is derived from a colorectal cancer tumor. The higher fold-enrichment of APC than beta-catenin is interesting and illustrates a potential limitation of this approach, as the enrichment of very low-abundance proteins may result in disproportionately high fold-enrichment values. For example, in several cases we observed known interaction partners that were not seen in the MS-based proteome profiling studies, making it impossible to calculate fold-enrichment for these proteins.
These examples illustrate some of the benefits of antibody target verification with IP-MS. The IP-MS approach uniquely verifies antibody capture performance by directly identifying peptide sequences from their putative targets. Beyond merely identifying the presence or absence of an antibody’s target, the IP-MS workflow enables the characterization of antibody selectivity by identifying the other proteins which are present in a sample following IP. This adds to the power of the overall method by identifying both off-target proteins and observing the presence of potential interacting partners which bind to antibody targets. Compared to other methods which characterize protein-protein interactions, the IP-MS approach is unique for its ability to filter and calculate protein enrichment. Further, IP-MS is capable of identifying target proteins, off-targets, and interactors in their native states (requiring no N- or C-terminal tags) and expression levels in biologically relevant cell lines. In the future, these workflows and analyses may be further extended to calculate fold-enrichment at the peptide level to assess the specificity of antibodies to targeted post-translational modification sites (e.g., phosphorylation, ubiquitination, and acetylation). Antibody selectivity measurements could also be used to help map antibody epitopes, protein conformations, or proteoforms that may have distinct protein interactions. These measurements could also help expedite the selection of complementary antibodies that may be combined in sandwich-type assays.
Unless otherwise stated, products identified with catalog numbers (Cat. No.) are from Thermo Fisher Scientific.
All cell lines were purchased from ATCC and grown in the condition noted in Table 1. All media and cell growth products were purchased from Thermo Fisher Scientific, including trypsin (Cat. Cells were grown to ~80% confluency and at passage 12–18 before lysis with IP Lysis Buffer (Cat. If cells underwent stimulation, cells were starved in 0.1% charcoal-stripped FBS (Cat. No. SH30068.01) for 24 hours before stimulation with 100 ng/mL of hIGF (Cell Signaling Technology Cat. No. 8917SF) or 100 nM insulin (Tocris Bioscience, Cat. No. 12585014) for 15 min and then lysed immediately. Protein concentration was determined by with the Pierce BCA Protein Assay Kit (Cat.
Cell model | Tissue type | 培地 | Media Cat. 番号 | Insulin | Stimulation |
---|---|---|---|---|---|
HCT116 | Colon | McCoy’s 5A | 16600-082 | N/A | ± IGF |
A549 | Lung | Hamm’s F12K | 21127-022 | N/A | ± IGF |
MCF7 | Breast | DMEM | 11995-040 | 10 µg/mL | N/A |
HepG2 | Liver | MEM | 11095-072 | N/A | ± Insulin |
LNCaP | Prostate | RPMI-1640 | 11875-085 | N/A | ± IGF |
NIH3T3 | Fibroblast | DMEM | 11995-040 | N/A | N/A |
BT-549 | Breast | RPMI-1640 | 11875-085 | 0.023I U/mL | N/A |
SK MEL 5 | Skin | DMEM | 11995-040 | N/A | N/A |
Hs 578T | Breast | DMEM | 11995-040 | 10 µg/mL | N/A |
SR | Lymphoblast | RPMI-1640 | 11875-085 | N/A | N/A |
HeLa | Cervical | DMEM | 11995-040 | N/A | N/A |
HEK293 | Kidney | DMEM | 11995-040 | N/A | N/A |
200–800 µg of lysate was further processed for analysis by mass spectrometry using the Pierce Mass Spec Sample Prep Kit for Cultured Cells (Cat. After the final drying step, samples were reconstituted in 0.1% trifluoroacetic acid (TFA) and cleaned of incompatible salts, detergents, and other reagents using the Pierce High pH Reverse-Phase Peptide Fractionation Kit (Cat. Samples were dried in a vacuum concentrator and reconstituted in 200 µL of 0.1% TFA. 5 µL of sample in 45 µL of water (1:5 dilution) was aliquoted and the Pierce Quantitative Fluorometric Peptide Assay (Cat.
For fractionation, 100 µg of digested peptide sample was fractionated with the Pierce High pH Reverse-Phase Peptide Fractionation Kit (Cat.
Fraction | Acetonitrile % | Acetonitrile (100%) µL | Triethylamine (0.1%) µL |
---|---|---|---|
1 | 5.0% | 50 | 950 |
2 | 6.25% | 62.5 | 937.5 |
3 | 7.5% | 75 | 925 |
4 | 8.75% | 87.5 | 912.5 |
5 | 10.0% | 100 | 900 |
6 | 15.0% | 150 | 850 |
7 | 20.0% | 200 | 800 |
8 | 50.0% | 500 | 500 |
Fractionated samples were dried in a vacuum concentrator and reconstituted in 20 µL of 2% acetonitrile and 0.1% formic acid. Peptide concentration was measured with the Pierce Quantitative Fluorometric Peptide Assay (Cat. Unfractionated and fractionated samples were transferred into an autosampler vial for LC-MS analysis.
2 µg of unfractionated and fractionated samples were analyzed by nanoLC-MS/MS on a Thermo Scientific Dionex UltiMate 3000 RSLCnano System and Thermo Scientific Q Exactive HF Hybrid Quadrupole-Orbitrap Mass Spectrometer using a Thermo Scientific EASY-Spray column (50 cm x 75 µm ID, PepMap C18, 2 µm particles, 100 Å pore size, Cat. No. ES803). The column temperature was maintained at 60°C using an Easy Spray Ion Source (Cat. No. ES081) interfaced online with the mass spectrometer. Mobile phase A (0.1% Formic acid in water, LC-MS grade) and Mobile phase B (0.1% Formic acid in Acetonitrile (ACN), LC-MS grade) were used to buffer the pH in the two running buffers. The total gradient was 210 min followed by a 30 min washout and re-equilibration. In detail, the flow rate started at 300 nL/min and 2% ACN with a linear increase to 20% ACN over 170 min followed by a 40 min linear increase to 32% ACN. The washout followed with a flow rate set to 400 nL/min at 95% ACN for 4 min followed by a 24 min re-equilibration at 2% ACN.
The Q Exactive HF instrument (located in Bremen, Germany) was freshly cleaned and calibrated using Tune (version 2.5 build 2042) instrument control software. Spray voltage was set to 1.9 kV, S-lens RF level at 60, and heated capillary at 275°C. Full scan resolutions were set to 120,000 at m/z 200. Mass range was set to 400–1,600. Isolation width was set at 2.0 Th. The normalized collision energy was set at 27. Peptide match was set to preferred, and isotope exclusion was utilized. All data was acquired in profile mode using positive polarity.
The Pierce MS-Compatible Magnetic IP Kit (Protein A/G) (Cat. No. 90409) was used to screen and verify antibodies as described in the instruction manual. 500 µg lysate and 3 µg or recommended dilutions of antibody were used for all experiments. IP eluates were dried in a vacuum concentrator and samples were spiked with green fluorescent protein (GFP) as a digestion indicator and then processed by an in-solution digestion method as recommended in the instruction manual for Cat. No. 90409. Dried digested samples were resuspended in 13 µL of 4% acetonitrile and 0.2% formic acid and transferred into autosampler vials before LC-MS Analysis.
The IP-MS samples were analyzed by nanoLC-MS/MS using a Thermo Scientific Dionex UltiMate 3000 RSLCnano System coupled to a Thermo Scientific Q Exactive HF Hybrid Quadrupole-Orbitrap Mass Spectrometer or Thermo Scientific Q Exactive Plus Orbitrap Mass Spectrometer. 7 µL of tryptic digest samples were desalted on-line using the Nano Trap Column (100 µm i.d. x 2 cm, packed with Acclaim PepMap100 C18, 5 µm, 100 Å, Cat. No. 164564), and separated using an EASY-Spray PepMap C18 column (15 cm x 75 µm ID, 3 µm particles, 100 Å pore size, Cat. No. ES800), with a total gradient time of 62 min. In detail, the flow rate started at 300 nL/min and 3% ACN with a linear increase to 25% ACN over 55 min followed by a 7 min linear increase to 40% ACN. The column was washed with a flow rate set to 600 nL/min at 95% ACN for 3 min followed by a 5 min re-equilibration at 3% ACN.
The Q Exactive HF and Q Exactive Plus instruments (located in Bremen, Germany) were freshly cleaned and calibrated. Spray voltage was set to 1.9 kV, S-lens RF level at 60, and heated capillary at 275°C. Full scan resolutions were set to 70,000 at m/z 200 (Q Exactive Plus) and 60,000 at m/z 200 (Q Exactive HF). The mass ranges for both instruments were set to 400–1,600 m/z. Instrument isolation widths were set at 1.2 Th for Q Exactive HF and 2.0 Th for Q Exactive Plus. The normalized collision energy was set at 27 for both instruments. Peptide match was set to preferred, and isotope exclusion was utilized. All data was acquired in profile mode using positive polarity.
RNA expression Z-scores were retrieved from CellMiner, hierarchically clustered with Cluster 3.0, and displayed with Java TreeView 1.16r4.
MS data obtained from unfractionated lysate, fractionated lysates, and deep proteome analysis and initial IP sample screens were analyzed using Proteome Discoverer 1.4 (release 1.14). A custom human proteome database (UniProt, assembled Feb 2016) was utilized for deep proteome analysis, while a combined database of human proteome and mouse/rat/rabbit IgGs, (UniProt, assembled July 2016) was used for database search IP screens. The IP database also included the recombinant protein A/G and GFP protein sequences. Trypsin was selected as the enzyme used for digestion. During automated searching, concatenated target/decoy databases were generated to validate peptide-spectral matches (PSMs) and filter identifications to a 1% false discovery rate (FDR). MS spectra were searched using 20 ppm precursor mass tolerance and 0.03 Da fragment tolerance. The data was searched with a static modification of carbamidomethylation of cysteine residues, and dynamic modifications including the acetylation of protein N-termini, oxidation of methionine residues, and phosphorylation of serine, threonine, and tyrosine residues.
Protein groups of unfractionated, fractionated, and IP-MS samples data were exported and custom software was used to extract the unique peptide sequences, number of PSMs, and top 3 peptide peak areas for each identified protein. Top 3 peptide peak area was used to determine relative abundance of specific proteins across multiple cell lines.
Fractionated proteome data was curated to determine the total number of proteins identified from each cell line. Specific protein targets were compared between cell lines using custom software to extract the number of peptide spectral matches (PSMs), unique peptide sequences, and both summed and averaged peptide peak areas or peptide intensities. Cell lines were selected for IP using these metrics to determine cell lines which expressed protein targets at a moderate abundance within 2 standard deviations of the mean protein intensity.
After testing multiple antibodies for the same target with replicates, IP data was first searched and screened using PD 1.4 using a combined database of the human proteome and mouse/rat/rabbit IgGs in addition to recombinant protein A/G and GFP sequences. The number of unique peptides, top 3 highest peptide intensities, PSMs, and total background proteins observed in each IP were assessed in order to verify the performance of each antibody to isolate its putative target.
Following PD 1.4 analysis, samples with detectable target were searched using MaxQuant 1.5.3.51 to obtain relative quantification of peptides and proteins and compare these protein abundances from replicate IP samples to unfractionated and fractionated proteome lysate samples. Curated contaminant proteins were also added to the database search. A target-decoy database was generated during automated searching and resulting peptide and protein identifications were filtered to a 1% FDR. Data was searched using group-specific parameters with a multiplicity of one, trypsin as the enzyme used for digestion, a maximum of 2 missed cleavages, fixed modification of carbamidomethylation of cysteine residues, and variable modifications including the acetylation of protein N-termini and oxidation of methionine residues. Label-free quantification (LFQ) was performed using a minimum LFQ ratio count of 2 and fast LFQ. Spectra were searched using a 20 ppm first search peptide tolerance and a 4.5 ppm main search peptide tolerance. MS/MS spectra were analyzed with a 20 ppm fragment match tolerance. Protein quantification was defined using a minimum threshold of 2 ratios, using unique and razor peptides for quantification. Large LFQ values were stabilized and required MS/MS for LFQ comparisons.
Once MaxQuant output was obtained, the data was manually analyzed to compare the intensities and LFQ values obtained across the unfractionated, fractionated, and replicate IP samples. Protein LFQ values were used to generate scatterplots to characterize the specificity of antibodies used in IP. For these scatterplots, LFQ values were plotted to compare the relative abundances of proteins identified in a “test” IP (plotted on the y-axis) to those proteins identified in a negative control IP for an unrelated target was un (plotted on the x-axis). The negative control antibody was selected for comparison either because the antibody recognized a different target or did not identify the target that was pulled down by the test IP. Plotting the relative abundances of proteins from each test and negative IP led to three distinct regions of the scatterplot, where proteins identified in both IPs were considered nonspecific “background” proteins along the diagonal of the scatterplot. Those proteins uniquely identified in the test IP were observed as aligned along the y-axis, while those proteins identified only in the negative control IP were aligned along the x-axis of the plot. Proteins observed uniquely in the test IP were color-coded according to their fold-enrichment versus deep proteome samples. Fold-enrichment and scatterplot calculations were performed by a custom web application to streamline the data analysis and generation of graphs for IP verification. LFQ values for replicate IPs were utilized to further filter the data for those proteins which were observed reproducibly across replicates. A 25% CV cutoff was used to filter proteins which were not reproducibly identified or quantified across replicate IP samples.
To calculate fold enrichment, for each MS run searched, the LFQ abundance of each protein was extracted and divided by the summed abundance of all proteins identified to obtain a “fraction” of that protein’s relative abundance versus every other protein identified in the sample. The relative fraction of the protein’s abundance in an IP sample was then compared to the fraction of the protein in the deep proteome samples to observe whether this fraction increased, decreased, or stayed the same relative to the other proteins that were identified in each IP. In this way, a fold-enrichment was calculated for every protein in the IP samples, and this calculation was used to characterize the enrichment of putative antibody targets and known target-protein interactors. These fold-enrichment calculations were performed using both protein LFQ.
Protein interactions were selected against the Homo sapiens proteome. Proteins were plotted according to their known interactors using text mining, experimental verification, database annotation, co-expression, gene fusion, and co-occurrence data. Data was plotted with nodes representing proteins uniquely identified in the test IP and edges representing evidence of protein-protein interactions. Protein fold-enrichment bar charts were color coded according to whether the identified protein was the putative antibody target or listed as a direct interactor with the target via the STRING database. Proteins were also color coded to represent whether they were indirect interactors (i.e., listed as interacting with annotated target interactors) or were not listed as interacting via the STRING database. Network statistics from the STRING database were downloaded with enriched GO terms for cellular component, biological processes, molecular function, KEGG pathways, Pfam annotations, and InterPro classifications.