Biotin and desthiobiotin labeling reagents are used for biotinylation of proteins, peptides, antibodies, and other biomolecules. These reagents target specific functional groups or residues on biomolecules, including primary amines and sulfhydryls. Biotin labeling reagents are offered as single-package reagents or convenient kits, as no-weigh packaging formats for quick and convenient working solutions, and as premium grade reagents with high-quality formulations.
Use the Biotinylation Reagent Selection Tool to aid in identifying the optimal labeling reagent for your application. Additionally, the Bioconjugation Technical Handbook can be used to support your biotin labeling experiments. Explore our Biotin Quantitation Reagents and HABA Calculator Tool to measure the amount of biotin attached to your biomolecule of interest.
What is biotinylation?
Biotinylation is the labeling of biotin to a molecule of interest such as a protein, antibody, peptide, or other biomolecule (Figure 1). Biotinylated proteins typically retain biological activity because the biotin group is relatively small. Biotin (Figure 2) is a naturally occurring vitamin that binds to avidin and streptavidin proteins with high affinity. The bond formation between biotin and avidin is rapid and, once formed, is unaffected by most extremes of pH, organic solvents, and other denaturing agents. Learn more about avidin-biotin interaction.
Biotin labeling reagents target specific functional groups or residues on biomolecules such as primary amines, sulfhydryls, carboxyls, or carbohydrates. Biotinylated proteins can be captured or detected by enzyme-linked immunosorbent assay (ELISA), western blot, immunohistochemistry (IHC), immunoprecipitation (IP), or flow cytometry applications. Learn more about biotinylation.
Choose the right biotin labeling reagent for your application using our Biotinylation Reagent Selection Tool, or select from our biotinylation reagent guide, our Biotin no-weigh packaging, or our Biotin premium grade reagents options below.
Figure 1. Biotinylated protein. If drawn to scale, the oval representing the protein would be many times larger than the structures and would likely contain several amino groups. Note that NHS is a leaving group (byproduct) in the reaction. The leaving group and any non-reacted biotin reagent are removed during the desalting step.
Figure 2. Biotin structure. Biotin, also known as B-vitamin B7 (formerly vitamin H and coenzyme R) is water soluble. The molecule is comprised of a ureido ring joined with a tetrahydrothiophene ring. A coenzyme for carboxylase enzymes, biotin is required for the synthesis of fatty acids, isoleucine and valine. Biotin is also involved in gluconeogenesis.
Biotinylation reagents based on reactive chemistry
*1-10 mg biotinylation kits include reagents for a HABA assay for measuring the level of biotin incorporation
*1-10 mg biotinylation kits include reagents for a HABA assay for measuring the level of biotin incorporation
Biotin no-weigh packaging
Biotin no-weigh packaging formats offer quick and convenient ready-to-use solutions. This format eliminates the need to weigh out small amounts of dry compounds and eliminates the waste of unused material. Simply reconstitute and the reagent is ready to use at the desired concentration.
A35358, A35390, A35389, A39256, A39257, A39258, A39259, A39260, A39261
Biotin premium grade reagents
Biotin premium grade reagents are high-quality reagents formulated for applications where product integrity and risk minimalization are essential. These premium grade reagents are ideal for long-term research projects that require consistent performance. Biotin premium grade products provide quality assurance review, lot sample retention, change control notification, along with an extra level of analytical testing and chemical characterization.
Comparison of specifications between premium and standard grade reagents
Test parameter | Test method | Premium grade | Standard grade |
---|
Purity | Quantitative NMR using an internal standard | Yes | Yes |
Visual | Color assessment | Yes | Yes |
Solubility | Sample dissolves at specified concentration in deionized water to yield a clear, colorless solution | Yes | Yes |
Identity | Infrared spectroscopy | Yes | Yes |
Mass identity | Mass spectrometry | Yes | NA |
Water content | Karl Fischer titration | Yes | NA |
Trace metals | Inductively coupled plasma mass spectrometry (ICP-MS) | Yes | NA |
Elemental analysis | Combustion analysis (values reported for C, H, N, O, and S) | Yes | NA |
Residual solvent analysis | Headspace gas chromatography | Upon request | NA |
Desthiobiotin vs. biotin
Desthiobiotin is a modified form of biotin that binds less tightly to avidin and streptavidin than biotin while still providing excellent specificity in affinity purification methods (Figure 3). Unlike biomolecules that are labeled with biotin, proteins and other targets that are labeled with desthiobiotin can be eluted without harsh, denaturing conditions. As a result, desthiobioinylation is an alternative to biotinylation for applications requiring gentle elution and recovery of the labeled target.
Figure 3. Comparison of the chemical structures of biotin and desthiobiotin.
16129, A39262, A39263, A39264, A39265
Assays and kits that use desthiobiotin
Product | Desthiobiotin reagent | Application |
---|
Desthiobiotinylation and Pull-Down Kit | Sulfo-NHS-LC-Desthiobiotin | Label a purified protein and use it to affinity purify interaction complexes in which it participates |
RNA 3' End Desthiobiotinylation Kit | Desthiobiotinylated Cytidine Bisphosphate | Enzymatically label an RNA oligo for use in Magnetic RNA-Protein Pull-Down Kit |
Active Serine Hydrolase Probes | Desthiobiotin-fluorophosphonate | Covalently label the active site of serine hydrolases to enable their selective enrichment |
Kinase Enrichment Kits | Desthiobiotin-ATP; Desthiobiotin-ADP | Covalently label the active site of ATPases, including chaperones and metabolic enzymes, to enable their selective enrichment |
Free download
Bioconjugation and crosslinking technical handbook
Everything you need to bioconjugate, crosslink, biotinylate, and modify proteins and peptides.
Download
Free download
Avidin-Biotin technical handbook
Everything you need to biotinylate, purify, or detect proteins.
Download
대량(bulk) 또는 맞춤형 제품 요청
단백질 라벨링, 교차결합 또는 변형 시약이 대량으로 필요하십니까? 대량 또는 맞춤형 크기로 제품을 제공합니다.
견적 요청