Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TopAir Systems is a manufacturer of innovative, safe, and power-saving clean-air and containment solutions. TopAir clean-air solutions are used in laboratories and manufacturing facilities within chemical/biological plants, universities, research and development facilities, and hospitals, as well as in the electronics, semiconductor, and pharmaceutical industries.
TopAir cabinets are known for their welded structures made of polypropylene, which is a superior anti-corrosive material able to protect staff from harsh acids which regular metal cabinets may not be able to withstand.
In addition to regular fume hoods, TopAir Systems also offer intelligent, active fume cabinets, high-quality PCR cabinets, balance enclosures, laminar flow clean benches, and ductless fume hoods with advanced safety features to protect your staff.
TopAir products can be customized to any size to suit your needs. Request a quote or get in contact with one of our specialists to find the right clean-air solution for you or browse from our featured options below.
Ductless fume hoods offer flexibility and mobility without sacrificing on safety and quality. TopAir Polypropylene Ductless Fume Hood PRO offers innovative safety features including a red-light alarm indicator, chemical sensor alarm to detect filter failure and saturation, and filter replacement reminders.
The Variable Air Volume system (VAV) varies the air velocity depending on sash opening and adjusts the internal fan speed accordingly to save energy, reduce noise, and improve cabinet performance.
Fume hoods protect laboratory staff from noxious fumes released by acids, dangerous gases, and organic solutions by channelling chemical vapours out of the building using an external fan installed on the roof or on an external wall.
TopAir Active Polypropylene Fume Hoods are a high-quality system enhanced with innovative technology. The fume hood features a sensor which detects staff standing next to the unit and opens and closes the sash automatically. Variable airflow technology allows the unit’s fan speed to adjust according to the sash position, reducing energy consumption.
Upon a failure, the whole interior of the cabinet lights up with a red light, so that staff, including people with hearing issues, can easily detect the failure from a distance.
PCR-UV + HEPA cabinets use an ozone-free UV lightbulb, as well as HEPA technology, to provide complete protection for your samples from contamination. The cabinet also offers a user-friendly 5″ control panel with UV lighting control and UV timer as well as a smart safety mechanism to prevent UV exposure. TopAir PCR cabinets are used for genomics, proteomics, molecular biology, and forensic sciences applications.
Horizontal and vertical-flow clean benches are used worldwide to provide product or sample protection for applications that do not require personal or environmental protection.
TopAir clean benches are designed to supply a clean controlled work environment meeting Class 100/ISO5 cleanliness standard, resembling a clean room, with the additional advantages of portability and small dimensions. TopAir provides a high-quality alternative to a clean room at a much lower cost and without any additional utility requirements.
Brochure
Specification sheets