Search Thermo Fisher Scientific
The Thermo Scientific Phenom Pharos G2 FEG-SEM brings field emission SEM to your tabletop. The Phenom Pharos G2 FEG-SEM will outperform many floor-standing SEMs in terms of image quality, while offering a vastly better user experience. For academic and industrial laboratories that so far did not consider SEM a realistic option, the Phenom Pharos G2 FEG-SEM makes FEG performance accessible thanks to its attractive form factor and short training required. Blazing fast sample loading means fast sample exchange, which means higher productivity. Unlike other SEMs, which end up being fully booked, the Phenom Pharos G2 FEG-SEM performs imaging and analysis jobs so quickly that it serves well as a walk-up tool.
The new Phenom Pharos G2 FEG-SEM expands its acceleration voltage range down to 1 kV, to better accommodate insulating and beam-sensitive samples, and up to 20 kV, with a resolution of 2.0 nm that reveals the finest details.
Introducing the Phenom Pharos G2 Desktop FEG-SEM: Intuitive evidence for your research
Unique among desktop SEMs, the Phenom Pharos G2 FEG-SEM offers a field emission source, which guarantees high brightness, crisp images, and stable beam current.
The Phenom Pharos G2 FEG-SEM offers a resolution of 2.0 nm at 20 kV. Such performance shows the shape of nanoparticles, imperfections in coatings, or other features that would be missed by tungsten SEMs or other tabletop SEMs.
With a voltage range down to 1 kV, the Phenom Pharos G2 FEG-SEM enables imaging of beam-sensitive samples, such as polymers, as well as insulating samples, without the requirement to apply a coating. As a result, nanoscale surface features are not obscured.
While FEG SEMs have a reputation for being difficult to accommodate and difficult to operate, the Phenom Pharos G2 FEG-SEM literally requires only a desk, and less than one hour of training. Master students, visitors, or other researchers typically not trained to work on high-end FEG SEMs can easily use the Phenom Pharos G2 FEG-SEM to create eye-catching images.
On the Phenom Pharos G2 FEG-SEM, morphological information is acquired together with compositional information, thanks to SE, BSE, and EDS detectors built into the system. A range of sample holders is available for temperature-controlled or electrical experiments.
Resolution |
|
Electron optical magnification range |
|
Light optical magnification |
|
Acceleration voltages |
|
Vacuum modes |
|
Detector |
|
Sample size |
|
Sample height |
|
This on-demand webinar has been designed to help you decide which SEM best meets your unique needs. We present an overview of Thermo Fisher Scientific SEM technology for multi-user research labs and focus on how these wide-ranging solutions deliver performance, versatility, in situ dynamics and faster time to results. Watch this webinar if you are interested in:
This on-demand webinar has been designed to help you decide which SEM best meets your unique needs. We present an overview of Thermo Fisher Scientific SEM technology for multi-user research labs and focus on how these wide-ranging solutions deliver performance, versatility, in situ dynamics and faster time to results. Watch this webinar if you are interested in:
A indústria moderna exige alta produtividade com qualidade superior, um equilíbrio mantido por meio de um controle de processo robusto. As ferramentas SEM e TEM com software de automação dedicado proporcionam informações rápidas e em várias escalas para monitoramento e aprimoramento de processos.
O controle de qualidade e a garantia de qualidade são essenciais na indústria moderna. Oferecemos uma gama de ferramentas de microscopia eletrônica e espectroscopia para análises multidimensionais e multimodais de defeitos, permitindo que você tome decisões confiáveis e informadas para controle e melhoria de processos.
Novos materiais são investigados em escalas cada vez menores para o máximo controle de suas propriedades físicas e químicas. A microscopia eletrônica fornece aos pesquisadores percepções importantes sobre uma ampla variedade de características materiais em escala micro a nano.
A espectroscopia por energia dispersiva (EDS) coleta informações elementares detalhadas juntamente com imagens de microscopia eletrônica, fornecendo contexto de composição crítico para observações EM. Com a EDS, a composição química pode ser determinada a partir de varreduras de superfície rápidas e holísticas que chegam até a átomos individuais.
A EDS proporciona informações de composição vitais para observações em microscópio eletrônico. Principalmente, nossos exclusivos sistemas detectores Super-X e Dual-X adicionam opções para maior produtividade e/ou sensibilidade, permitindo otimizar a aquisição de dados para atender às prioridades de pesquisa.
A pesquisa de materiais modernos depende cada vez mais da análise nanométrica em três dimensões. A caracterização 3D, incluindo dados de composição para contexto químico e estrutural completos, é possível com a EM 3D e a espectroscopia de raios X por energia dispersiva.
A EDS de resolução atômica fornece um contexto químico sem igual para análise de materiais, diferenciando a identidade elementar de átomos individuais. Quando combinado com a TEM de alta resolução, é possível observar a organização precisa dos átomos em uma amostra.
O estudo de materiais em condições reais muitas vezes envolve trabalhar em altas temperaturas. O comportamento dos materiais à medida que eles se recristalizam, derretem, deformam ou reagem na presença de calor pode ser estudado in situ com microscopia eletrônica de varredura ou ferramentas DualBeam.
A observação direta e em tempo real de alterações microestruturais com a microscopia eletrônica é necessária para entender os princípios subjacentes de processos dinâmicos, como recristalização, crescimento de grãos e transformação de fases durante o aquecimento, o resfriamento e a umidificação.
Os materiais novos precisam ser analisados em resolução cada vez maior, mantendo o contexto maior da amostra. A análise em várias escalas permite correlacionar várias ferramentas e modalidades de geração de imagens, como a microTC de raios X, DualBeam, laser PFIB, SEM e TEM.
A espectroscopia por energia dispersiva (EDS) coleta informações elementares detalhadas juntamente com imagens de microscopia eletrônica, fornecendo contexto de composição crítico para observações EM. Com a EDS, a composição química pode ser determinada a partir de varreduras de superfície rápidas e holísticas que chegam até a átomos individuais.
A EDS proporciona informações de composição vitais para observações em microscópio eletrônico. Principalmente, nossos exclusivos sistemas detectores Super-X e Dual-X adicionam opções para maior produtividade e/ou sensibilidade, permitindo otimizar a aquisição de dados para atender às prioridades de pesquisa.
A pesquisa de materiais modernos depende cada vez mais da análise nanométrica em três dimensões. A caracterização 3D, incluindo dados de composição para contexto químico e estrutural completos, é possível com a EM 3D e a espectroscopia de raios X por energia dispersiva.
A EDS de resolução atômica fornece um contexto químico sem igual para análise de materiais, diferenciando a identidade elementar de átomos individuais. Quando combinado com a TEM de alta resolução, é possível observar a organização precisa dos átomos em uma amostra.
O estudo de materiais em condições reais muitas vezes envolve trabalhar em altas temperaturas. O comportamento dos materiais à medida que eles se recristalizam, derretem, deformam ou reagem na presença de calor pode ser estudado in situ com microscopia eletrônica de varredura ou ferramentas DualBeam.
A observação direta e em tempo real de alterações microestruturais com a microscopia eletrônica é necessária para entender os princípios subjacentes de processos dinâmicos, como recristalização, crescimento de grãos e transformação de fases durante o aquecimento, o resfriamento e a umidificação.
Os materiais novos precisam ser analisados em resolução cada vez maior, mantendo o contexto maior da amostra. A análise em várias escalas permite correlacionar várias ferramentas e modalidades de geração de imagens, como a microTC de raios X, DualBeam, laser PFIB, SEM e TEM.
Para garantir o desempenho ideal do sistema, fornecemos acesso a uma rede de especialistas em serviços de campo, suporte técnico e peças de reposição certificadas.