SEM EDS elemental analysis
Experience the next level of EDS analysis with Thermo Scientific™ ChemiSEM™ Technology, a game-changing integration of SEM and EDS functions in a single, seamless user interface. Unleash the power of elemental information with speed, accuracy, and unparalleled simplicity.
ChemiSEM Technology user interface
What is ChemiSEM Technology?
ChemiSEM™ Technology, is an advanced and revolutionary solution designed to address the complex needs of customers in material characterization. It caters to the curious minds seeking to understand the composition of intricate materials, unravel the secrets of their samples, and detect even the tiniest impurities.
ChemiSEM brings together live quantification and decades of EDS analysis expertise to deliver elemental information quickly and effortlessly. With ChemiSEM™, you get reliable results, uncover hidden features, and access complete data, all at your fingertips.
ChemiSEM EDS elemental analysis
ChemiPhase image of a refractory material (brown fused alumina) employed in steel production. The phase analysis shows the location of the alumina (light blue) but also the presence of unexpected materials that act as contamination, such as titanium oxide (displayed in pink) and fragments of silica (displayed in yellow)
Advantages of SEM EDS analysis with ChemiSEM Technology
Always On
Say goodbye to moving between SEM imaging and EDS analysis for each sample. ChemiSEM™ continuously collects the EDS signal during SEM imaging, capturing a full EDS spectrum at every point in the image.
Faster Data Acquisition
Access elemental information faster than ever before with innovative data segmentation techniques based on machine learning and ultrafast signal processing. Seamlessly integrated with SEM, ChemiSEM™ streamlines the entire process.
Reliable Results
Enjoy accurate qualification and quantification across a wide range of operating conditions, including beam energy, sample size, and working distance. ChemiSEM™ ensures consistent and dependable outcomes.
Complete Information
See the big picture with comprehensive micro-scale elemental composition. Immediate color results highlight defects or imperfections that might have been missed with conventional techniques.
Multi-Data Viewing
Get a complete characterization of your sample at a glance with simultaneous viewing of Secondary Electron Detector (SED) and elemental information in a single frame.
Simplified Operation
ChemiSEM™ makes elemental information accessible to everyone. Its intuitive user interface allows easy access to compositional results, enabling more scientists and engineers to utilize your facilities effectively.
Phase analysis with ChemiSEM Technology
Introducing ChemiPhase, the game-changing feature in ChemiSEM that's revolutionizing phase analysis. Imagine pinpointing unique material phases effortlessly. With ChemiPhase's advanced big data approach, detecting statistically significant spectra within your datacube is a breeze.
Complex materials analysis
Characterizing diverse phases has never been easier. ChemiPhase, the unbiased statistical engine, leaves no room for error, avoiding unexpected element misses due to overlaps or intensities.
Experience the Benefits:
  • Automated Brilliance: Say goodbye to user bias. ChemiPhase handles the heavy lifting without prior element identification.
  • Pixel-Perfect Precision: Identify major and minor components down to a single pixel, free from peak overlaps.
  • Speed and Reliability: Achieve phase determination in under a minute, even from minimal X-ray data (just 10 counts per pixel).
  • Reduced Complexity: Navigate with confidence using image-spectrum pairs for each distinct phase, while spectra are optimized automatically.
Elevate your material analysis game with ChemiPhase!
Thermo Scientific SEM instruments featuring ChemiSEM Technology:
Discover the capabilities of ChemiSEM™ Technology with our cutting-edge products
Phenom Desktop SEM
Axia ChemiSEM
Apreo 2 SEM
Quattro ESEM
ChemiSEM Technology is always on and enables any user to acquire elemental data—you will have more access to complete information than ever before. See just a few examples of the utility of simultaneous EDS and SEM analysis in the videos below.
Elemental information in the blink of an eye
This video is taken on the Thermo Scientific Prisma E SEM equipped with a 30 mm2 EDS detector. We demonstrate the speed of ChemiSEM Technology with composition automatically identified (AutoID) and easy selection of elements of interest.
Highlight contrast that would otherwise go unnoticed
ChemiSEM Technology is always on – you no longer have to switch applications when you need elemental information; instead live results are shown in the SEM user interface. This video highlights several forms of elemental information (point and area).
Reliable results
Elemental information is useless when it is not reliable. This video shows how ChemiSEM Technology is capable of dealing with X-ray peak overlap and deconvolution – making sure that your elemental information is correct and relevant, even in live color imaging.
Resources
BioLab Axia Customer Testimonial
BioLab Axia Customer Testimonial
Applications
Quality Control_Thumb_274x180_144DPI
Controle de qualidade
O controle de qualidade e a garantia de qualidade são essenciais na indústria moderna. Oferecemos uma gama de ferramentas de microscopia eletrônica e espectroscopia para análises multidimensionais e multimodais de defeitos, permitindo que você tome decisões confiáveis e informadas para controle e melhoria de processos.
Fundamental Materials Research_R&D_Thumb_274x180_144DPI
Pesquisa de materiais fundamentais
Novos materiais são investigados em escalas cada vez menores para o máximo controle de suas propriedades físicas e químicas. A microscopia eletrônica fornece aos pesquisadores percepções importantes sobre uma ampla variedade de características materiais em escala micro a nano.
Samples
Pesquisa de baterias
O desenvolvimento de baterias tornou-se possível pela análise em várias escalas com microTC, SEM e TEM, espectroscopia Raman, XPS e visualização e análise 3D digital. Saiba como essa abordagem fornece as informações estruturais e químicas necessárias para construir baterias melhores.
Saiba mais ›
Pesquisa de polímeros
A microestrutura de polímero determina as características e o desempenho do material em massa. A microscopia eletrônica permite uma análise abrangente micrométrica da morfologia e da composição do polímero para aplicações de Pesquisa e Desenvolvimento e de controle de qualidade.
Saiba mais ›
Pesquisa de metais
A produção efetiva de metais requer um controle preciso de inclusões e precipitações. Nossas ferramentas automatizadas podem executar uma série de tarefas críticas para a análise de metais, incluindo: contagem de nanopartículas, análise química de EDS e preparação de amostras de TEM.
Saiba mais ›
Forênsica
Os microtraços de evidências de cena de crime podem ser analisados e comparados usando a microscopia eletrônica como parte de uma investigação forense. As amostras compatíveis incluem fragmentos de vidro e de tinta, marcas de ferramentas, drogas, explosivos e resíduo de disparo de arma de fogo.
Saiba mais ›
Petróleo e gás
À medida que a demanda por petróleo e gás continua, há uma necessidade contínua de extração eficiente e eficaz de hidrocarbonetos. A Thermo Fisher Scientific oferece uma variedade de soluções de microscopia e espectroscopia para diferentes aplicações de ciência do petróleo.
Saiba mais ›
Pesquisa geológica
A geociência depende da observação consistente e precisa em várias escalas de recursos em amostras de rochas. A SEM-EDS, combinada com software de automação, permite a análise direta e em larga escala da textura e da composição mineral para a pesquisa da petrologia e da mineralogia.
Saiba mais ›
Teste de materiais automotivos
Cada componente em um veículo moderno é projetado para segurança, eficiência e desempenho. A caracterização detalhada dos materiais automotivos pela microscopia eletrônica e pela espectroscopia informa as decisões críticas do processo, as melhorias do produto e os novos materiais.
Saiba mais ›
Fibras e filtros
O diâmetro, a morfologia e a densidade das fibras sintéticas são parâmetros fundamentais que determinam a vida útil e a funcionalidade de um filtro. A microscopia eletrônica de varredura (SEM) é a técnica ideal para investigar essas características de forma rápida e fácil.
Saiba mais ›
Products
Folha de estilo para cartões de instrumentos original
Axia ChemiSEM
  • Mapeamento elementar quantitativo ao vivo
  • Imagens por microscopia eletrônica de varredura de alta fidelidade
  • Flexível e fácil de usar, mesmo para usuários iniciantes
  • Manutenção fácil
Prisma E SEM
  • SEM de nível básico com excelente qualidade de imagem
  • Carregamento e navegação de amostras fáceis e rápidos para várias amostras
  • Compatível com uma ampla gama de materiais graças aos modos de vácuo dedicados
Quattro ESEM
  • FEG SEM de alta resolução ultraversátil com capacidade ambiental única (ESEM)
  • Observe todas as informações de todas as amostras com imagens simultâneas SE e BSE em cada modo de funcionamento
Apreo 2 SEM
  • SEM de alto desempenho para resolução geral nanométrica ou subnanométrica
  • Detector retroespelhado T1 na coluna para contraste de materiais sensíveis e com taxa de TV
  • Excelente desempenho em longas distâncias de trabalho (10 mm)
Phenom Pharos Desktop SEM
  • Fonte FEG com faixa de tensão de aceleração de 2 kV a 15 kV
  • Resolução de <2,5 nm (SE) e <4,0 nm (BSE) a 15 kV; ampliação de até 1.000.000 vezes
  • Detector EDS e SE opcional totalmente integrado
Phenom XL G2 Desktop SEM
  • Para amostras grandes (100 x 100 mm) e ideal para automação
  • Resolução <10 nm e ampliação de até 200.000 vezes; tensão de aceleração de 4,8 kV até 20 kV
  • Detector EDS e BSE opcional totalmente integrado
Phenom ProX Desktop SEM
  • SEM de mesa de alto desempenho com detector EDS integrado
  • Resolução <8 nm (SE) e <10 nm (BSE); ampliação de até 150.000 vezes
  • Detector SE opcional
Phenom Pro Desktop SEM
  • SEM de mesa de alto desempenho
  • Resolução <8 nm (SE) e <10 nm (BSE); ampliação de até 150.000 vezes
  • Detector SE opcional
Phenom Pure Desktop SEM
  • SEM de mesa de nível básico
  • Resolução <25 nm; ampliação de até 65.000 vezes
  • Fonte CeB6 de longa duração
Phenom Perception GSR Desktop SEM
  • GSR de mesa SEM automatizado dedicado
  • Resolução <10 nm; ampliação de até 200.000 vezes
  • Fonte CeB6 de longa duração
Phenom ParticleX AM Desktop SEM
  • SEM de mesa versátil com software de automação para fabricação aditiva
  • Resolução <10 nm; ampliação de até 200.000 vezes
  • Detector SE opcional
Phenom ParticleX TC Desktop SEM
  • Desktop SEM versátil com software de automação para limpeza técnica
  • Resolução <10 nm; ampliação de até 200.000 vezes
  • Detector SE opcional
Phenom ParticleX Steel Desktop SEM
  • SEM e EDS integrados
  • Fácil de usar
  • Inclusões submicrométricas
Style Sheet for Komodo Tabs
Style Sheet to change H2 style to p with em-h2-header class
Contact us
Ready to revolutionize your materials characterization? Contact us now to learn more about ChemiSEM™ Technology and explore how it can elevate your research and analysis to new heights.