Search Thermo Fisher Scientific
最も多く使用されている溶媒や溶媒ブレンドには、LC-MSグレードの蒸留水やアセトニトリル、トリフルオロ酢酸(TFA)やギ酸(FA)、ヘプタフルオロ酪酸(HFBA)などのイオンペア試薬などがあります。これらの溶媒は、単一要素の処方液としても、あるいは便利なブレンドとしてもご使用いただけます。溶媒は、ISO9001に準拠した工場で生産されており、分析証明書と一緒にご提供します。溶媒と溶媒ブレンドは、Thermo Scientific™ 質量分析計を使用してバリデートされています。
Trifluoroacetic Acid (TFA) | Formic Acid (FA) | 0.1% TFA in Acetonitrile, LC-MS Grade | 0.1%TFA in Water, LC-MS Grade | 0.1% FA in Acetonitrile, LC-MS Grade | 0.1% FA in Water, LC-MS Grade | |
Structure | ||||||
Formulation | Solvent | Solvent | Solvent Blend | Solvent Blend | Solvent Blend | Solvent Blend |
Formats | Liquid (in ampules or bottles) | Liquid (in ampules or bottles) | Liquid (bottles) | Liquid (bottles) | Liquid (bottles) | Liquid (bottles) |
Specifications measured | >10 | 7-20 | >20 | >20 | >20 | >20 |
Order now | Order now | Order now | Order now | Order now | Order now |
アセトニトリルに混合したThermo Scientific™ Pierce™ 0.1%トリフルオロ酢酸の仕様。