Search Thermo Fisher Scientific
こうしたプロテアーゼ/ホスファターゼ混合型の阻害剤は、セリン、スレオニン、チロシンホスファターゼを標的とするとともに、セリン、システイン、アスパラギン酸プロテアーゼ、およびアミノペプチダーゼも阻害します。金属プロテアーゼはEDTAの添加により阻害されます。EDTAは個別のバイアル入りの液体タイプと、タブレットタイプの両方に含まれています。こうした混合型の阻害剤は、タンパク質分解を防止すると同時に、リン酸化タンパクを維持するため、サンプル中のタンパクの完全な保護を可能にします。
Thermo Scientific™ Haltliquid cocktailsは、100 μLの1回使い切りタイプまたは1.5mLおよび10 mLサイズで入手可能です。Thermo Scientific Pierce tabletsは、10 mL用があります。様々な容量および価格のニーズに対応しています。
Halt Protease and Phosphatase Inhibitor Cocktail | Halt Protease and Phosphatase Inhibitor Cocktail, EDTA free | Pierce Protease and Phosphatase Inhibitor Tablet | Pierce Protease and Phosphatase Inhibitor Tablet, EDTA Free | |
Flexible addition based on sample volume? | Yes | Yes | No | No |
---|---|---|---|---|
Contains EDTA | Yes, but in a separate vial | No | Yes | No |
Contains DMSO | Yes | Yes | No | No |
Requires reconstitution | No (100X) | No (100X) | Yes | Yes |
Protein assay compatibility | All | All | BCA (dilute sample 1:3), | BCA (dilute sample 1:3) |
Downstream compatibility | Not compatible with 2D or IMAC | All | Not compatible with 2D or IMAC | All |
Product sizes available | 24 x 100 μL, 1 mL, 5 x 1 mL, 10 mL | 24 x 100 μL, 1 mL, 5 x 1 mL, 10 mL | 30 tablets | 30 tablets |
Order now | Order now | Order now | Order now |