Search Thermo Fisher Scientific
3 | 405 | 525/40 | 437 | 478 | (in buffer) 3 | flow cytometry |
Invitrogen Brilliant Violet™ 480 (BV480) is a tandem dye excited by the 405 nm violet laser and emits at 480 nm. This dye has a medium relative brightness and can be used for cell surface, intracellular, and nuclear staining.
When using two or more Super Bright, Brilliant Violet™, Brilliant Ultra Violet™, or other polymer dye- conjugated antibodies in a staining panel, we recommend using Super Bright Complete Staining Buffer or Brilliant Stain Buffer to minimize any non-specific polymer interactions.
We offer BV480 dye conjugated to primary antibodies for use in flow cytometry.
Spectral signature of Brilliant Violet™ 480 dye. Data acquired on a 5-laser Cytek Aurora and normal human peripheral blood cells stained with CD8a Monoclonal Antibody (RPA-T8), Brilliant Violet™ 480 were used for analysis.
Intracellular staining of mouse splenocytes using Brilliant Ultra Violet™ 480. C57BL/6 mouse splenocytes were unstimulated (left) or stimulated for 48 hours with CD3e Monoclonal Antibody, Functional Grade (right). Cells were then surface-stained with CD45R (B220) Monoclonal Antibody, FITC and stained intracellularly, using the Foxp3/Transcription Factor Staining Buffer Set and protocol, with 1.0 µg of Ki-67 Monoclonal Antibody (SolA15), Brilliant Violet™ 480. Viable cells in the lymphocyte gate were used for analysis, as determined by LIVE/DEAD™ Fixable Blue Dead Cell Stain Kit, for UV excitation.
Cell surface staining of mouse splenocytes using Brilliant Violet™ 480. C57BL/6 mouse splenocytes were stained with CD4 Monoclonal Antibody, FITC and 0.25 µg of Rat IgG2a kappa Isotype Control, Brilliant Violet™ 480 (left) or 0.25 µg of CD8a Monoclonal Antibody, Brilliant Violet™ 480 (right). Viable cells in the lymphocyte gate were used for analysis, as determined by 7-AAD Viability Staining Solution.
Brilliant Ultra Violet dyes are a polymer dye-based technology and compatible with both spectral flow cytometry as well as traditional flow cytometry. The UV laser has unique channels far from heavily occupied detectors, allowing for larger panels.
Protocols that fit your needs in flow cytometry ranging from sample preparation to numerous cell stimulation conditions, staining, immunophenotyping, and data analysis strategies.
A handy reference poster featuring the broad range of our available dyes and labeling reagents.
Protocols that fit your needs in imaging ranging from sample and assay preparation to staining, labeling, and data analysis strategies.
A tool for selecting the optimal fluorescent dyes for your Invitrogen EVOS cell imaging systems.
Cy™ is a trademark or registered trademark of GE Healthcare.
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and are used under license.
For Research Use Only. Not for use in diagnostic procedures.
Brilliant Ultra Violet™ 和 Brilliant Violet™ 是 Becton, Dickinson 公司或其附属公司的商标或注册商标,经许可使用。
仅供科研使用,不可用于诊断目的。