Search Thermo Fisher Scientific
The Tali® Image-Based Cytometer can be used to detect green and red fluorescence of a variety of dyes and stains. This table ranks the fluorescence intensity of various dyes when examined on the Tali® Image-Based Cytometer. Unless otherwise indicated, cells were labeled with the indicated fluorescent dye or stain at concentrations suggested in the Product Information Sheet and analyzed on the Tali® Image-Based Cytometer using the “Green+Red Setting” setting. In some cases, there was spectral overlap of the fluorescence emission (commonly known as bleedthrough) into the opposite channel. This information is also included in the table.
Download the Application Note
Dye, Product Tested | Excitation (nm) | Emission (nm) | Expected Emission Channel | Green Channel Signal | Red Channel Signal | Recommendations |
---|---|---|---|---|---|---|
7-AAD (7-aminoactinomycin D) | 549 | 645 | Red |
None
|
+++
| Use 1-2 ul per ml. |
R-PE (R-phycoerythrin), CD3-RPE conjugate | 564 | 577 | Red |
None
|
++
| Test titration of antibody conjugate. |
CellROX Orange Reagent | 545 | 565 | Red | None | +++ | Use according to PIS. |
MitoTracker® Orange CM-H2TMRos | 551 | 575 | Red |
None
|
++
| Use according to PIS. |
Ethidium homodimer-1 | 528 | 618 | Red |
None
|
+++
| Use 1 uM. |
Rhodamine Red-X anti-mouse secondary | 573 | 591 | Red |
None
|
+
| basically undetectable |
Dye, Product Tested | Excitation (nm) | Emission (nm) | Expected Emission Channel | Green Channel Signal | Red Channel Signal | Recommendations |
---|---|---|---|---|---|---|
SYTOX® Green stain | 504 | 523 | Green |
+++
|
+
| Use 12.5 nM. |
LIVE/DEAD® Fixable Green Dead Cell Stain | 495 | 520 | Green |
+++
|
++
| Spectral overlap into red channel at lowest concentration tested (0.25 µL/mL) |
CellTrace™ Green | 492 | 517 | Green |
+++
|
+
| Use at 50 nM–250 nM |
CellEvent® Green reagent | 502 | 530 | Green |
++
|
None
| 10 µM final concentration |
SYTO® 9 stain | 485 | 498/501 | Green |
+++
|
None
| Use 100 nM |
TO-PRO®-1 | 515 | 531 | Green |
++
|
++
| Due to spectral overlap can not be used for multiplexing experiments |
FM® 1-43X | 510 | 626 | Green |
+++
|
+++
| Due to spectral overlap can not be used for multiplexing experiments |
CellTracker™ Green CFDA-AM | 492 | 517 | Green |
+++
|
None
| Use according to PIS. |
5-(and-6)-carboxy-2',7' -dichlorofluorescein diacetate (carboxy-DCFDA) *mixed isomers* | 505 | 525 | Green |
++
|
None
| Use at 25-50 uM. |
Annexin V-FITC | 494 | 518 | Green | ++ | None | Use according to PIS (test 10 ul Annexin V-FITC if signal is dim). |
Acridine Orange | 500 | 526 | Green | +++ | None | For Jurkat cells suggest using 0.25 – 0.5 uM (test titration of the dye to ensure no overlap into the red channel) |
Calcein AM | 494 | 517 | Green | +++ | + | Use at 0.1 - 1 uM (test titration of dye to ensure no overlap into red channel). |
"+" undetectable to barely detectable on the histogram, but can see spots on the Tali images.
"++" can detect peak on histogram, but may be difficult to separate positive and negative peaks.
"+++" easily detectable peaks on histogram. Positive and negative populations can be easily resolved.