Search Thermo Fisher Scientific
Find answers to frequently asked questions about the Tali® Image Cytometer such as basic technical aspects, physical features, computer compatibility and archiving, and cellular analysis capabilities.
How long does it take to run an assay on the Tali® Image Cytometer?
Assay Time (sec) | ||||||||
---|---|---|---|---|---|---|---|---|
Number of fields | Quick Count | Viability | Apoptosis | GFP | GFP + Viability | RFP | RFP + viability | GFP + RFP |
4 | 7 | 13 | 17 | 17 | 22 | 20 | 25 | 33 |
9 | 13 | 26 | 36 | 42 | 55 | 40 | 53 | 72 |
13 | 18 | 37 | 52 | 58 | 78 | 62 | 75 | 105 |
18 | 24 | 50 | 72 | 81 | 106 | 84 | 103 | 142 |
20 | 26 | 56 | 79 | 91 | 117 | 93 | 114 | 159 |
|
|
|
Product | Green Channel | Red Channel | For use with: | Cat. No. |
---|---|---|---|---|
Tali® Viability Kit—Dead Cell Green | SYTOX® Blue dye | N/A | RFP cells | T10787 |
Tali® Viability Kit—Dead Cell Red | N/A | Propidium iodide (PI) | GFP cells | T10786 |
Tali® Apoptosis Kit | Annexin V—Alexa Fluor® 488 dye | Propidium iodide (PI) | Apoptosis studies | T10788 |
Channel | Excitation | Emission | Use or Fluorophores Detected |
---|---|---|---|
Bright field | White LED | -- | Visualize and count cells |
Green | 466/40 nm (Blue LED) | 525/50 nm | Green fluorescent protein (GFP), Alexa Fluor® 488 dye, SYTOX® Blue dye |
Red | 543/22 nm (Green LED) | 585 LP | Red fluorescent protein (RFP), Propidium iodide (PI) |
What are the physical dimensions of the Tali® cytometer?
What slides are compatible with Tali® Image Cytometer?
Does Tali® Image Cytometer require a computer?
What cell types have been tested?
For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.