Find complete Tali ® specifications on this 3-channel (bright field, green fluorescence, red fluorescence) benchtop assay platform that performs suspension cell-based assays.
General Specifications
Physical
Characteristics
  • Instrument type: Benchtop cell analyzer and suspension cell-based assay platform
  • Instrument dimensions: 11 ½” (W) X 11 ½” (H) X 17 ½” (D)
  • Weight: 19.4 lbs (8.8 kg)
  • Operating power: 100–240 VAC, 2.5 A, 120 V
  • Frequency: 50/60 Hz
  • Electrical input: 12 VDC, 13 A
  • Installation site: Indoor use only
  • Operating temperature: 15–29°C
  • Operating humidity: < 80% (non-condensing)
Technical Specifications 
  • Processing time: 10 seconds to 2 minutes, depending on the number of fields captured and complexity of assay selected
  • Sample concentration range: 1 × 105–1 × 107 cells/mL
  • Recommended sample diameter range: 5–60 μm
  • Required sample volume: 25 μL
  • Firmware: Tali™ Image Cytometer Firmware (visit www.invitrogen.com/tali for updates)
  • USB Drive: 4 Gigabyte
Optics 
  • Optics: 3 channels (bright field, green fluorescence, red fluorescence)
  • Excitation: Green channel LED: 458 ± 20 nm
  • Red channel LED: 530 ± 20 nm
  • Filters: Green channel: 466/40 EX, 495 LP Di, 525/50 EM
  • Red channel: 543/22 EX, 580 LP Di, 585 LP EM
  • Camera: 1.3 Mega pixels, 4X objective, 4X or 16X digital zoom
Tali™ Cellular Analysis Slide 
  • Material: Polylactic acid (PLA)
  • Dimensions: 110 mm (w) × 24 mm (d) × 1.9 mm (h)
  • Chamber volume: 25 μL