Search Thermo Fisher Scientific
The Applied Biosystems™ QuantStudio™ 6 Flex Real-Time PCR System is ideal for laboratories with multiple applications and end users on a limited budget. With a planned upgrade path to a Applied Biosystems™ QuantStudio™ 7 Flex System that accommodates automation or TaqMan Array Cards, the QuantStudio 6 Flex System is an ideal qPCR platform to accommodate changing future needs.
Minimize up-front capital investment
With one instrument, you can interchange between 96-well, 96-well Fast, and 384-well formats.
Skip the learning curve
The intuitive software, easy touch screen setup, and effortless block changes are designed to let you get started right away.
Achieve sensitivity and performance
Detect as small as 1.5-fold changes in singleplex reactions and 10 logs dynamic range.
Upgrade when you need it
The QuantStudio 6 Flex system can be seamlessly upgraded to a QuantStudio 7 Flex system by a field service engineer to give you additional automation, throughput, and multiplexing capabilities.
QuantStudio 6 Flex | QuantStudio 7 Flex Learn more › | QuantStudio 12 Flex Learn more › | QuantStudio 3D Learn more › | |
---|---|---|---|---|
Colors | 5 | 6 (21 filter combinations) | 6 (21 filter combinations) | 2 (endpoint detection) |
Chemistry | SYBR®, TaqMan® | SYBR®, TaqMan® | SYBR®, TaqMan® | TaqMan® Only |
Available blocks | 96-well 96-well Fast 384-well | 96-well 96-well Fast 384-well TaqMan® Array Card (384-well microfluidic card) | 96-well 96-well Fast 384-well TaqMan® Array Card (384-well microfluidic card) OpenArray® plates (3,072 through-holes) | 20,000 partitions/chip |
Automation available | No | Yes | Yes | No |
Throughput | *** | **** | ***** | * |
Block change | Block change from front in less than 1 minute, no tools required | N/A | ||
21CFR Part 11 enablement | Optional software module available | |||
Remote monitoring | Available to monitor up to 15 networked instruments simultaneously | |||
Intuitive touch screen | Yes | Yes | Yes | Yes |
Key applications |
|
|
|
|
See product list below | Request a quote › | Request a quote › | Request a quote › |
The QuantStudio 6 Flex Real-Time PCR System is supported with:
Minimize up-front capital investment
With one instrument, you can interchange between 96-well, 96-well Fast, and 384-well formats.
Skip the learning curve
The intuitive software, easy touch screen setup, and effortless block changes are designed to let you get started right away.
Achieve sensitivity and performance
Detect as small as 1.5-fold changes in singleplex reactions and 10 logs dynamic range.
Upgrade when you need it
The QuantStudio 6 Flex system can be seamlessly upgraded to a QuantStudio 7 Flex system by a field service engineer to give you additional automation, throughput, and multiplexing capabilities.
QuantStudio 6 Flex | QuantStudio 7 Flex Learn more › | QuantStudio 12 Flex Learn more › | QuantStudio 3D Learn more › | |
---|---|---|---|---|
Colors | 5 | 6 (21 filter combinations) | 6 (21 filter combinations) | 2 (endpoint detection) |
Chemistry | SYBR®, TaqMan® | SYBR®, TaqMan® | SYBR®, TaqMan® | TaqMan® Only |
Available blocks | 96-well 96-well Fast 384-well | 96-well 96-well Fast 384-well TaqMan® Array Card (384-well microfluidic card) | 96-well 96-well Fast 384-well TaqMan® Array Card (384-well microfluidic card) OpenArray® plates (3,072 through-holes) | 20,000 partitions/chip |
Automation available | No | Yes | Yes | No |
Throughput | *** | **** | ***** | * |
Block change | Block change from front in less than 1 minute, no tools required | N/A | ||
21CFR Part 11 enablement | Optional software module available | |||
Remote monitoring | Available to monitor up to 15 networked instruments simultaneously | |||
Intuitive touch screen | Yes | Yes | Yes | Yes |
Key applications |
|
|
|
|
See product list below | Request a quote › | Request a quote › | Request a quote › |
The QuantStudio 6 Flex Real-Time PCR System is supported with:
Applied Biosystems™ analysis modules make it easier and faster to view, analyze, remotely access, store, and share qPCR data. Discover Standard Curve, Genotyping, and Relative Quantification analysis apps now.