Search Thermo Fisher Scientific
The PE-eFluor 610 dye is a high-performance tandem fluorophore designed for use with flow cytometers equipped with blue (488 nm), green (532 nm), or yellow-green (561 nm) laser. It has a peak emission of 607 nm that can be detected using typical filters for PE-Texas Red dye, such as a 610/20 bandpass filter.
Initial brightness | PE-eFluor 610 dye is a tandem fluorophore developed for use in multicolor flow cytometry. | ||
Photostability in buffer |
488/532/561 | 610/20 | 532-561 | 607 | ||
Laser line | Common filter set | Excitation max | Emission max |
PE-eFluor 610 dye is brighter than PE-Texas Red dye. Normal human peripheral blood cells were stained with CD3 conjugated to PE-eFluor 610 dye (clone UCHT1) (dark orange line) or PE-Texas Red dye (light orange line).
Photostability of PE-eFluor 610 dye. Normal human peripheral blood cells were stained with Anti-CD3 PE-eFluor 610 (clone UCHT1). Cells were then washed and incubated for six hours at 4°C in the dark (red line), at room temperature in ambient light (blue line), or at room temperature on the windowsill (orange line).
We offer PE-eFluor 610 dye conjugated to primary antibodies for use in flow cytometry.
See all PE-eFluor 610 Primary Antibodies
See all PE-eFluor 610 Secondary Antibodies
Brilliant Ultra Violet™ 和 Brilliant Violet™ 是 Becton, Dickinson 公司或其附属公司的商标或注册商标,经许可使用。
仅供科研使用,不可用于诊断目的。