PerCP-Cyanine5.5 dye
Peridinin chlorophyll protein-Cyanine5.5 (PerCP-Cyanine5.5) is a photostable tandem dye excited from the 488 nm laser. It is stable in fixative, and stained cells can be stored overnight. Less-abundant intracellular antigens stained with PerCP-Cyanine5.5 can often be reliably detected in multicolor flow cytometry experiments.
We offer PerCP-Cyanine5.5 dye conjugated to a variety of antibodies.
PerCP-Cyanine5.5 dye dashboard
Initial brightness
PerCP-Cyanine5.5 dye is a near-IR fluorophore used primarily for flow cytometry, optimally excited by the blue laser. 
Photostability in buffer
 Medium488695/40482690 
Relative staining index
Laser line
Common filter set
Excitation max
Emission max
PerCP-Cyanine5.5 dye products
We offer PerCp-Cyanine5.5 dye conjugated to a variety of antibodies and streptavidin conjugates.
Antibody conjugates
Streptavidin conjugates
Brilliant Ultra Violet™ 和 Brilliant Violet™ 是 Becton, Dickinson 公司或其附属公司的商标或注册商标,经许可使用。
仅供科研使用,不可用于诊断目的。