Search Thermo Fisher Scientific
Invitrogen eBioscience Super Bright 600 dye is tandem polymer dye, consisting of violet (405 nm) laser–excitable Super Bright 436 and an acceptor dye that emits fluorescence signal at 600 nm. The technology behind the Super Bright polymer dye family makes them useful for identifying low-abundance markers on a cell surface in flow cytometry because they emit fluorescence with high efficacy.
An alternative to Brilliant Violet 605 conjugates, antibody conjugates of Super Bright 600 provide increased resolution of positive and negative populations, but with less background and less interfering dye–dye interaction than Brilliant Violet 605 conjugates.
Executing a well-designed experiment requires multiple reagents. Super Bright antibody conjugates are fully compatible with Invitrogen UltraComp eBeads microspheres for compensation, viability stains, fluorescent dyes and other reagents commonly used in flow cytometry. Robust, highly sensitive, and stable, Super Bright conjugates easily fit into any flow cytometry panel.
We offer Super Bright 600 dye conjugated to primary antibodies for use in flow cytometry.
Initial brightness | The violet laser–excitable Super Bright 600 dye is optimized for use in flow cytometry applications. | ||
Photostability in buffer |
405 | 610/20 | 414 | 600 | ||
Laser line | Common filter set | Excitation max | Emission max |
We offer Super Bright 600 dye conjugated to primary antibodies for use in flow cytometry.
Invitrogen eBioscience Super Bright polymer dyes are violet laser–excitable fluorophores that expand the capabilities of multi-parametric flow cytometry analysis. This family of bright fluorophores enables the use of more markers off the violet channel and increases the detection of low-abundance antigens due to their super bright signals.
The Super Bright polymer dyes join the trusted Invitrogen Alexa Fluor dye and Invitrogen eFluor dye portfolios in providing fluorochromes compatible with the 405 nm spectral line of the violet laser.
Brilliant Ultra Violet™ 和 Brilliant Violet™ 是 Becton, Dickinson 公司或其附属公司的商标或注册商标,经许可使用。
仅供科研使用,不可用于诊断目的。