Protein detection and analysis—Western Blotting
Our all-encompassing range of solutions for western blotting analysis is meticulously designed and tested to meet the highest standards and minimize time and effort. Establish a robust foundation for your western blotting experiments.
Innovative Western Blotting Solutions
Boost your productivity with western blotting tools designed for efficient high-quality data generation. 
Everyday essentials for western blotting success
Limit the variables in your experiments by using dependable products from one trusted source.
Speed, performance and convenience
Total protein normalization
Total protein normalization (TPN) for quantitative western blotting analysis offers several advantages over traditional housekeeping protein-based (HKP) normalization. One optimal protein labeling reagent for TPN is the No-Stain Protein Labeling Reagent. TPN using No-Stain Labeling Reagent exhibits good linearity of response within the concentration range typically investigated (1–80 µg of protein load). The wider dynamic range prevents the loss of valuable quantitative data due to signal saturation, a common occurrence with HKP normalization. TPN using No-Stain Labeling Reagent simplifies experimental design, helping save time and resources. Unlike HKP normalization, there is no need to verify that the expression of housekeeping proteins remains unaffected by the treatment. No-Stain Labeling Reagent is particularly well-suited for TPN due to its rapid labeling protocol, and compatibility with downstream transfer and immunoblotting. It enables sensitive and stable signal detection down to 20 ng of proteins, allowing for accurate quantitation of protein expression levels.
Learn more about TPN with No-Stain Labeling Reagent
Multiplexed fluorescent western blotting
Multiplexed western blotting is a powerful technique that offers several advantages over traditional western blotting. Its ability to simultaneously detect and quantify multiple proteins within a single sample helps increase efficiency, improves accuracy, reduces costs, and enhances data quality. It is advantageous when investigating complex biological pathways involving proteins with diverse molecular weights. The simultaneous detection of multiple protein targets in one experiment reduces experimental variability and is an excellent choice when the sample volume is limited or when analyzing large sample sets. Fluorescent western blotting requires careful planning and experiment design. Selecting the right products from the start reduces optimization and troubleshooting time and helps ensure the success of your experiment.
Design your next multiplex western blotting experiment
Western blot learning and troubleshooting
Whether you are new to western blotting, or an experienced researcher wanting to confirm your knowledge, access our educational tips and resource materials to help with every aspect of western blot analysis.
Access western blotting learning center
video icon
Videos
Discover the new ways to do western blotting with our how-to videos.
clipboard with checkmark icon
Protocols
Find recipes for buffers, stock solutions, and general western blot protocols.
book catalog internal serach icon
Foundation Library
Tech tips, application notes, handbooks and more.
gears troubleshooting icon
Troubleshooting
Overcome western blot difficulties with our troubleshooting guidance.
Stylesheet for Classic Wide Template adjustments
Hero banner dark gradient overlay
웨스턴 블로팅은 중요한 단백질 생물학 도구입니다.
단백질 검출과 분석에 적합한 제품 조합을 찾아보세요. 웨스턴 블로팅 워크플로우 전 단계에 걸친 종합적인 솔루션을 통해 최소한의 시간과 노력으로 고품질의 결과를 도출할 수 있습니다.
웨스턴 블로팅 교육 자료
웨스턴 블로팅을 처음 진행하시는 분들이나 경험 있는 숙련된 연구자분들 모두에게 유용한 웨스턴 블로팅 교육 자료를 통해 웨스턴 블로팅 분석의 모든 측면에서 도움을 얻어보세요.
혁신적인 웨스턴 블로팅 솔루션
효율적으로 고품질 데이터를 생성할 수 있도록 설계된 웨스턴 블로팅 툴로 생산성을 높여보세요.
Invitrogen iBright 이미징 시스템
  • 강력한 9.1메가픽셀(MP) 카메라—논문 품질의 이미지 캡처 가능
  • 스마트 노출 포착 기술—최적의 노출 시간을 신속하게 판단
  • 단순한 인터페이스와 자동 기능 패키지—최소의 노력으로 신속하게 이미지 캡처
  • 멀티 애플리케이션 지원—단백질 겔, 핵산 겔, 화학발광 웨스턴 블롯, 형광 웨스턴 블롯 등
  • 최대 4개의 Fluorophores 다중 분석
  • 21 CFR Part 11 준수 지원
Invitrogen iBright 이미징 시스템
  • 강력한 9.1메가픽셀(MP) 카메라—논문 품질의 이미지 캡처 가능
  • 스마트 노출 포착 기술—최적의 노출 시간을 신속하게 판단
  • 단순한 인터페이스와 자동 기능 패키지—최소의 노력으로 신속하게 이미지 캡처
  • 멀티 애플리케이션 지원—단백질 겔, 핵산 겔, 화학발광 웨스턴 블롯, 형광 웨스턴 블롯 등
  • 최대 4개의 Fluorophores 다중 분석
  • 21 CFR Part 11 준수 지원
시작 단계에 도움이 되는 웨스턴 블로팅 툴
어디서부터 시작해야 할까요?
제품 가이드를 다운로드받고, 선정 가이드 및 교육센터를 통해 웨스턴 블로팅 성공에 필요한 모든 것을 찾아보세요.