c7c8 - 主要特点/性能数据/相关资源/应用/技术/文档/联系人
Helios 5 Laser PFIB System
The Thermo Scientific Helios 5 Laser PFIB System combines the best-in-class monochromated Elstar Scanning Electron Microscopy (SEM) Column with a plasma focused ion beam (PFIB) and a femtosecond laser to produce a high-resolution imaging and analysis tool with in-situ ablation capability, offering unprecedented material removal rates for fast millimeter-scale characterization at nanometer resolution.
Statistically relevant subsurface and 3D characterization
The femtosecond laser can cut many materials at rates orders of magnitude faster than a typical gallium FIB; a large (100s of micrometers) cross-section can be created within 5 minutes. As the laser has a different removal mechanism (ablation vs the ion sputtering of FIB), it can easily process challenging materials, such as non-conductive or ion-beam sensitive samples.
High-quality large volume analysis
The extremely short duration of the femtosecond laser pulses introduces almost no artifacts such as heat impact, microcracking, melting, or those typical of traditional mechanical polishing. In most cases, the laser-milled surfaces are clean enough for direct SEM imaging and even for surface-sensitive techniques such as electron backscatter diffraction (EBSD) mapping.
Built on the proven Helios 5 PFIB platform, this instrument incorporates a suite of state-of-the-art technologies to provide high-performance, high-resolution transmission electron microscopy (TEM) and atom probe tomography (APT) sample preparation and extremely high-resolution SEM imaging with precise materials contrast.
Key Features
Fast material removal
Millimeter-scale cross sections with up to 15,000x faster material removal than a typical gallium focused ion beam.
Statistically relevant subsurface and 3D data analysis
Acquire data for much larger volumes within a shorter amount of time.
Accurate and repeatable cut placement
The same coincident point for all 3 beams (SEM/PFIB/laser) enables accurate and repeatable cut placement and 3D characterization.
Fast characterization of deep subsurface features
Extraction of subsurface TEM lamella or chunks for 3D analysis.
High throughput processing of challenging materials
Includes non-conductive or ion-beam-sensitive samples.
Fast and easy characterization of air-sensitive samples
No need to transfer samples between different instruments for imaging and cross-sectioning.
Shares all capabilities of the Helios 5 PFIB platform
High-quality gallium-free TEM and APT sample preparation and high-resolution imaging capabilities.
Specifications
产品表格规范样式表
Femtosecond-laser specifications
Laser integration
  • Fully integrated in the chamber with the same coincident point for all 3 beams (SEM/PFIB/laser), enabling accurate and repeatable cut placement and 3D characterization.
Laser Output
First Harmonic
  • Wavelength
1030 nm (IR)
  • Pulse duration
<280 fs
Second Harmonic
  • Wavelength
515 nm (green)
  • Pulse duration
<300 fs
Optics 
Coincident point
  • Working distance= 4 mm (Same as SEM/FIB)
Objective lens
  • Variable (motorized)
Polarization
  • Horizontal/vertical
Repetition rate
  • 1 kHz – 1 MHz
Position accuracy
  • <250 nm
Protective shutter
  • Automated SEM/PFIB protective shutter
Software
  • Laser control software
  • Laser 3D serial sectioning workflow
  • Laser 3D serial sectioning workflow with EBSD
  • Laser scripting with optional Thermo Scientific AutoScript 4 Software
Safety
  • Interlocked laser enclosure (Class 1 laser safety)
Style Sheet for Komodo Tabs
Resources
On demand webinar: Discover the all new Helios 5 Laser PFIB
Register for our recorded webinar and learn how the combination of fs-Laser and PFIB provides mm-scale subsurface and 3D analysis at nm resolution and enables new workflows such as fast characterization of air sensitive samples and multi-scale correlative microscopy with deep subsurface sample extraction. 
Register now
Latest application developments of multiple ion species plasma FIB technology
Register for our live webinar and learn how leading research labs are using our new Thermo Scientific Helios 5 Laser PFIB and Thermo Scientific Helios 5 Hydra DualBeam to advance their materials characterization.
Register now
On demand webinar: Discover the all new Helios 5 Laser PFIB
Register for our recorded webinar and learn how the combination of fs-Laser and PFIB provides mm-scale subsurface and 3D analysis at nm resolution and enables new workflows such as fast characterization of air sensitive samples and multi-scale correlative microscopy with deep subsurface sample extraction. 
Register now
Latest application developments of multiple ion species plasma FIB technology
Register for our live webinar and learn how leading research labs are using our new Thermo Scientific Helios 5 Laser PFIB and Thermo Scientific Helios 5 Hydra DualBeam to advance their materials characterization.
Register now
Applications
采用电子显微镜进行过程控制
采用电镜进行过程控制
现代工业需求高通量、质量卓越、通过稳健的工艺控制维持平衡。SEM扫描电镜TEM透射电镜工具结合专用的自动化软件,为过程监控和改进提供了快速、多尺度的信息。
使用电子显微镜进行质量控制和故障分析
质量控制和故障分析
质量控制和保证对于现代工业至关重要。我们提供一系列用于缺陷多尺度和多模式分析的 EM电子显微镜和光谱工具,使您可以为过程控制和改进做出可靠、明智的决策。
使用电镜进行基础材料研究
基础材料研究
越来越小的规模研究新型材料,以最大限度地控制其物理和化学特性。电子显微镜为研究人员提供了对微米到纳米级各种材料特性的重要见解。
Techniques
3D 材料表征
材料开发通常需要多尺度3D表征。DualBeam仪器可实现大体积连续切片和随后纳米级的SEM成像,可用于样品的高质量3D重建。
了解更多 ›
(S)TEM 样品制备
DualBeam显微镜可制备用于(S)TEM分析的高质量、超薄样品。借助先进的自动化技术,任何经验水平的用户都可以获得各种材料的专家级结果。
了解更多 ›
APT 样品制备
原子探针断层扫描 (APT) 提供材料的原子分辨率 3D 组成分析。聚焦离子束 (FIB) 显微镜是一项为 APT 表征进行高质量、定向和特定样品制备的基本技术。
了解更多 ›
交叉切片
交叉切片通过揭示亚表面信息提供额外的剖析。DualBeam仪器配备卓越的聚焦离子束色谱柱,可实现高质量的交叉切片。借助自动化技术,可实现无人参与的高通量样品处理功能。
了解更多 ›
原位实验
需要通过电子显微镜直接实时观察微观结构变化,以便了解在加热、冷却和润湿过程中的动态过程(如再结晶、晶粒生长和相变)的基本原理。
了解更多 ›
多尺度分析
必须在更高分辨率下分析新材料,同时保留较大的样品背景。多尺度分析允许多种成像工具和模态(如X射线microCT、DualBeam、激光PFIB、SEM和TEM)关联。
了解更多 ›
3D 材料表征
材料开发通常需要多尺度3D表征。DualBeam仪器可实现大体积连续切片和随后纳米级的SEM成像,可用于样品的高质量3D重建。
了解更多 ›
(S)TEM 样品制备
DualBeam显微镜可制备用于(S)TEM分析的高质量、超薄样品。借助先进的自动化技术,任何经验水平的用户都可以获得各种材料的专家级结果。
了解更多 ›
APT 样品制备
原子探针断层扫描 (APT) 提供材料的原子分辨率 3D 组成分析。聚焦离子束 (FIB) 显微镜是一项为 APT 表征进行高质量、定向和特定样品制备的基本技术。
了解更多 ›
交叉切片
交叉切片通过揭示亚表面信息提供额外的剖析。DualBeam仪器配备卓越的聚焦离子束色谱柱,可实现高质量的交叉切片。借助自动化技术,可实现无人参与的高通量样品处理功能。
了解更多 ›
原位实验
需要通过电子显微镜直接实时观察微观结构变化,以便了解在加热、冷却和润湿过程中的动态过程(如再结晶、晶粒生长和相变)的基本原理。
了解更多 ›
多尺度分析
必须在更高分辨率下分析新材料,同时保留较大的样品背景。多尺度分析允许多种成像工具和模态(如X射线microCT、DualBeam、激光PFIB、SEM和TEM)关联。
了解更多 ›
用于将 H2 样式更改为具有 em-h2-header 类 p 的样式表