Search Thermo Fisher Scientific
当社はほぼすべての電気泳動解析を単純化しスピードアップできる、便利でコスト効率が高いプレキャストゲルおよびゲルシステムをご提供します。
Order | Order | Order | Order | Order | Order | |
Quickly analyze 48-96 proteins | Analyze post-translational protein modifications | Excellent resolution for low MW | Optimized resolution of active proteases | Superior resolution of native proteins | Convenient & simplified 2D electrophoresis | |
---|---|---|---|---|---|---|
E-PAGE™ Gels | Novex® IEF Gels | Novex® Tricine Gels | Novex® Zymogram Gels | NativePAGE® Gels | ZOOM® 2D Electrophoresis System | |
Applications | Compatible with high-throughput systems | Separate native proteins & study modifications | Separation of low MW proteins & peptides | Separation of proteases based on size & activity | Determine size & purity of native proteins | Fractionate proteins before electrophoresis & MS |
Separation range | 10–200 kDa | pl 3.5–6.8 pl 3.5–8.5 | 2–20 kDa | 10–220 kDa | 15–10,000 kDa | pH 3–12 |
Max sample volume/well | 20 µL (E-PAGE™ 48) 15 µL (E-PAGE™ 96) | 20 µL | 25 µL | 20 µL | 25 µL | 140 µL (diluted) |
Gel % available | 8% (E-PAGE™ 48) 6% (E-PAGE™ 96) | 5% | 10%, 16%, 10–20% | 10% (with gelatin) 12% & 4–16% (with casein) | 3–12%, 4–16% | pH 3–10 NL, 3–10L, 4–7, 6–10, 9–12 (broad range) pH 4.5–5.5, 5.3–6.3, 6.1–7.1 (narrow range) |
Average run time | 14 min | 2.5 hr | 90 min | 90 min | 90 min | 30 min (1st dimension) 40 min (2nd dimension) |
Shelf life | 6 months | 2 months | 1–2 months | 2 months | 6 months | 12 months |
Gel dimensions | 13.5 x 10.8 cm, 3.7 mm thick | 8 x 8 cm, 1.0 mm thick | 8 x 8 cm, 1.0 mm thick | 8 x 8 cm, 1.0 mm thick | 8 x 8 cm, 1.0 mm thick | 7 cm strips, 0.5 cm thick |